Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96132357-96132586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTL
Entrez Gene ID
431705
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Gene Aliases
SAS1B
UniGene
Hs.447993
Gene Chromosome Location
Chr.2: 96122817-96143059 on Build GRCh38
Transcripts
NM_001002036.3 XM_011511205.2 XM_011511206.2 XM_011511207.2 XM_011511208.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934011 c.592G>T p.D198Y Chr.2: 96132585-96132585 on Build GRCh38
227435 c.625G>A p.E209K Chr.2: 96132552-96132552 on Build GRCh38
6542837 c.610C>A p.R204S Chr.2: 96132567-96132567 on Build GRCh38
6606117 c.622A>T p.N208Y Chr.2: 96132555-96132555 on Build GRCh38
5863007 c.592G>A p.D198N Chr.2: 96132585-96132585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764030100 Chr.2: 96132506-96132506 on Build GRCh38
rs1014150375 Chr.2: 96132554-96132554 on Build GRCh38
rs752138502 Chr.2: 96132517-96132517 on Build GRCh38
rs976753473 Chr.2: 96132469-96132469 on Build GRCh38
rs768248187 Chr.2: 96132532-96132532 on Build GRCh38
rs141578566 Chr.2: 96132400-96132400 on Build GRCh38
rs752897981 Chr.2: 96132499-96132499 on Build GRCh38
rs957721103 Chr.2: 96132443-96132443 on Build GRCh38
rs367836103 Chr.2: 96132473-96132473 on Build GRCh38
rs764556124 Chr.2: 96132560-96132560 on Build GRCh38
rs765548772 Chr.2: 96132493-96132493 on Build GRCh38
rs573922224 Chr.2: 96132474-96132474 on Build GRCh38
rs541100535 Chr.2: 96132521-96132521 on Build GRCh38
rs201337537 Chr.2: 96132549-96132549 on Build GRCh38
rs553274942 Chr.2: 96132435-96132435 on Build GRCh38
rs779697688 Chr.2: 96132526-96132526 on Build GRCh38
rs1024066340 Chr.2: 96132393-96132393 on Build GRCh38
rs757683346 Chr.2: 96132519-96132519 on Build GRCh38
rs115782187 Chr.2: 96132490-96132490 on Build GRCh38
rs370913603 Chr.2: 96132527-96132527 on Build GRCh38
rs996165744 Chr.2: 96132559-96132559 on Build GRCh38
rs927719468 Chr.2: 96132404-96132404 on Build GRCh38
rs748161965 Chr.2: 96132552-96132552 on Build GRCh38
rs41320144 Chr.2: 96132566-96132566 on Build GRCh38
rs1055209461 Chr.2: 96132446-96132446 on Build GRCh38
rs143172084 Chr.2: 96132567-96132567 on Build GRCh38
rs910545878 Chr.2: 96132449-96132449 on Build GRCh38
rs930932621 Chr.2: 96132488-96132488 on Build GRCh38
rs756407081 Chr.2: 96132524-96132524 on Build GRCh38
rs944942846 Chr.2: 96132464-96132464 on Build GRCh38
rs377277711 Chr.2: 96132489-96132489 on Build GRCh38
rs772118993 Chr.2: 96132553-96132553 on Build GRCh38
rs376251705 Chr.2: 96132505-96132505 on Build GRCh38
rs746228775 Chr.2: 96132523-96132523 on Build GRCh38

View Full Product Details