Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88448665-88448896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXI3
Entrez Gene ID
344167
Gene Name
forkhead box I3
Gene Aliases
-
UniGene
Hs.716722
Gene Chromosome Location
Chr.2: 88448207-88452535 on Build GRCh38
Transcripts
NM_001135649.1

SNP Detail

SNP ID Chromosome Location
rs563411808 Chr.2: 88448724-88448724 on Build GRCh38
rs768084842 Chr.2: 88448795-88448795 on Build GRCh38
rs760093211 Chr.2: 88448778-88448778 on Build GRCh38
rs544018503 Chr.2: 88448706-88448706 on Build GRCh38
rs764032108 Chr.2: 88448749-88448749 on Build GRCh38
rs562206043 Chr.2: 88448698-88448698 on Build GRCh38
rs763315727 Chr.2: 88448727-88448727 on Build GRCh38
rs1012230746 Chr.2: 88448687-88448687 on Build GRCh38
rs923448937 Chr.2: 88448757-88448757 on Build GRCh38
rs770906850 Chr.2: 88448716-88448716 on Build GRCh38
rs187343162 Chr.2: 88448837-88448837 on Build GRCh38
rs371562127 Chr.2: 88448847-88448847 on Build GRCh38
rs926696421 Chr.2: 88448719-88448719 on Build GRCh38
rs75621379 Chr.2: 88448715-88448715 on Build GRCh38
rs867424861 Chr.2: 88448685-88448685 on Build GRCh38
rs756470601 Chr.2: 88448824-88448824 on Build GRCh38
rs754678456 Chr.2: 88448865-88448865 on Build GRCh38
rs976182686 Chr.2: 88448733-88448733 on Build GRCh38
rs558453364 Chr.2: 88448688-88448688 on Build GRCh38
rs4972212 Chr.2: 88448871-88448871 on Build GRCh38
rs1051942259 Chr.2: 88448802-88448802 on Build GRCh38
rs564911510 Chr.2: 88448699-88448699 on Build GRCh38
rs772775131 Chr.2: 88448801-88448801 on Build GRCh38
rs753462098 Chr.2: 88448848-88448848 on Build GRCh38
rs942057193 Chr.2: 88448762-88448762 on Build GRCh38
rs955268781 Chr.2: 88448768-88448768 on Build GRCh38
rs182321240 Chr.2: 88448763-88448763 on Build GRCh38
rs542613834 Chr.2: 88448794-88448794 on Build GRCh38
rs910859728 Chr.2: 88448834-88448834 on Build GRCh38

View Full Product Details