Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32403060-32403285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL2
Entrez Gene ID
56244
Gene Name
butyrophilin like 2
Gene Aliases
BTL-II, BTN7, HSBLMHC1, SS2
UniGene
Hs.534471
Gene Chromosome Location
Chr.6: 32393339-32408879 on Build GRCh38
Transcripts
NM_001304561.1 XM_017011057.1 NR_136245.1

Gene Symbol
LOC101929163
Entrez Gene ID
101929163
Gene Name
uncharacterized LOC101929163
Gene Aliases
-
UniGene
Hs.57692
Gene Chromosome Location
Chr.6: 32255173-32407730 on Build GRCh38
Transcripts
NM_001304561.1 XM_017011057.1 NR_136245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3921482 c.510C>T p.F170F Chr.6: 32403134-32403134 on Build GRCh38
249151 c.451delC p.H151fs*96 Chr.6: 32403193-32403193 on Build GRCh38
4003976 c.542G>A p.R181Q Chr.6: 32403102-32403102 on Build GRCh38
4797662 c.543G>A p.R181R Chr.6: 32403101-32403101 on Build GRCh38
5870313 c.456_457GG>AA p.M152_E153>IK Chr.6: 32403187-32403188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771818649 Chr.6: 32403103-32403103 on Build GRCh38
rs774396428 Chr.6: 32403129-32403129 on Build GRCh38
rs754302847 Chr.6: 32403089-32403089 on Build GRCh38
rs1041686572 Chr.6: 32403172-32403172 on Build GRCh38
rs143181335 Chr.6: 32403170-32403170 on Build GRCh38
rs138358964 Chr.6: 32403210-32403210 on Build GRCh38
rs57116766 Chr.6: 32403209-32403209 on Build GRCh38
rs141591068 Chr.6: 32403211-32403211 on Build GRCh38
rs770703868 Chr.6: 32403111-32403111 on Build GRCh38
rs759516191 Chr.6: 32403121-32403121 on Build GRCh38
rs746683370 Chr.6: 32403107-32403107 on Build GRCh38
rs60740710 Chr.6: 32403193-32403193 on Build GRCh38
rs28362681 Chr.6: 32403102-32403102 on Build GRCh38
rs778551969 Chr.6: 32403101-32403101 on Build GRCh38
rs769681867 Chr.6: 32403123-32403123 on Build GRCh38
rs753198266 Chr.6: 32403251-32403251 on Build GRCh38
rs766759467 Chr.6: 32403159-32403159 on Build GRCh38
rs573958434 Chr.6: 32403193-32403194 on Build GRCh38
rs1015448256 Chr.6: 32403246-32403246 on Build GRCh38
rs142404581 Chr.6: 32403080-32403080 on Build GRCh38
rs200373630 Chr.6: 32403202-32403202 on Build GRCh38
rs745713767 Chr.6: 32403099-32403099 on Build GRCh38
rs756449026 Chr.6: 32403086-32403086 on Build GRCh38
rs59129682 Chr.6: 32403131-32403131 on Build GRCh38
rs746606560 Chr.6: 32403185-32403185 on Build GRCh38
rs75415018 Chr.6: 32403083-32403083 on Build GRCh38
rs371561394 Chr.6: 32403226-32403226 on Build GRCh38
rs60263670 Chr.6: 32403198-32403198 on Build GRCh38
rs546466362 Chr.6: 32403149-32403149 on Build GRCh38
rs757932339 Chr.6: 32403184-32403184 on Build GRCh38
rs35935572 Chr.6: 32403263-32403264 on Build GRCh38
rs754211446 Chr.6: 32403161-32403161 on Build GRCh38
rs370253771 Chr.6: 32403194-32403194 on Build GRCh38
rs9461742 Chr.6: 32403082-32403082 on Build GRCh38
rs775371318 Chr.6: 32403203-32403203 on Build GRCh38
rs777527157 Chr.6: 32403106-32403106 on Build GRCh38
rs765719298 Chr.6: 32403242-32403242 on Build GRCh38
rs767602775 Chr.6: 32403137-32403137 on Build GRCh38
rs115653647 Chr.6: 32403216-32403216 on Build GRCh38
rs78587369 Chr.6: 32403150-32403150 on Build GRCh38
rs777022452 Chr.6: 32403221-32403221 on Build GRCh38
rs781030325 Chr.6: 32403197-32403197 on Build GRCh38
rs776296960 Chr.6: 32403116-32403116 on Build GRCh38
rs149101528 Chr.6: 32403169-32403169 on Build GRCh38
rs770558436 Chr.6: 32403190-32403190 on Build GRCh38
rs755501406 Chr.6: 32403099-32403099 on Build GRCh38
rs138875868 Chr.6: 32403183-32403183 on Build GRCh38

View Full Product Details