Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183445456-183445717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN2AIP
Entrez Gene ID
55602
Gene Name
CDKN2A interacting protein
Gene Aliases
CARF
UniGene
Hs.592508
Gene Chromosome Location
Chr.4: 183444591-183449064 on Build GRCh38
Transcripts
NM_001317343.1 NM_017632.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
481091 c.295A>G p.K99E Chr.4: 183445557-183445557 on Build GRCh38
1725015 c.383_384insA p.G129fs*5 Chr.4: 183445645-183445646 on Build GRCh38
3730333 c.343T>G p.Y115D Chr.4: 183445605-183445605 on Build GRCh38
1053610 c.358G>A p.E120K Chr.4: 183445620-183445620 on Build GRCh38
1053609 c.274T>C p.Y92H Chr.4: 183445536-183445536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551218956 Chr.4: 183445603-183445603 on Build GRCh38
rs774669140 Chr.4: 183445485-183445485 on Build GRCh38
rs964035681 Chr.4: 183445618-183445618 on Build GRCh38
rs1000956513 Chr.4: 183445483-183445483 on Build GRCh38
rs748434981 Chr.4: 183445554-183445554 on Build GRCh38
rs779381798 Chr.4: 183445562-183445562 on Build GRCh38
rs774924031 Chr.4: 183445574-183445574 on Build GRCh38
rs562786754 Chr.4: 183445591-183445591 on Build GRCh38
rs397713406 Chr.4: 183445481-183445482 on Build GRCh38
rs749682911 Chr.4: 183445567-183445567 on Build GRCh38
rs760306455 Chr.4: 183445510-183445510 on Build GRCh38
rs531711240 Chr.4: 183445592-183445592 on Build GRCh38
rs775189235 Chr.4: 183445509-183445509 on Build GRCh38
rs1012159416 Chr.4: 183445524-183445524 on Build GRCh38
rs764939439 Chr.4: 183445611-183445611 on Build GRCh38
rs141073811 Chr.4: 183445558-183445558 on Build GRCh38
rs576993790 Chr.4: 183445516-183445516 on Build GRCh38
rs778181086 Chr.4: 183445513-183445513 on Build GRCh38
rs200183773 Chr.4: 183445602-183445602 on Build GRCh38
rs756211383 Chr.4: 183445682-183445682 on Build GRCh38
rs771140793 Chr.4: 183445569-183445569 on Build GRCh38
rs11282266 Chr.4: 183445476-183445477 on Build GRCh38
rs1052348592 Chr.4: 183445504-183445505 on Build GRCh38
rs369246349 Chr.4: 183445543-183445543 on Build GRCh38
rs146932991 Chr.4: 183445655-183445655 on Build GRCh38
rs369471031 Chr.4: 183445503-183445503 on Build GRCh38
rs542248144 Chr.4: 183445607-183445607 on Build GRCh38
rs201633995 Chr.4: 183445476-183445476 on Build GRCh38
rs35858568 Chr.4: 183445477-183445478 on Build GRCh38
rs752852591 Chr.4: 183445676-183445676 on Build GRCh38
rs776091820 Chr.4: 183445609-183445609 on Build GRCh38
rs761674330 Chr.4: 183445610-183445610 on Build GRCh38
rs376417441 Chr.4: 183445523-183445523 on Build GRCh38
rs750436686 Chr.4: 183445623-183445623 on Build GRCh38
rs753566867 Chr.4: 183445608-183445610 on Build GRCh38
rs766870406 Chr.4: 183445507-183445507 on Build GRCh38
rs73870523 Chr.4: 183445522-183445522 on Build GRCh38
rs758251245 Chr.4: 183445649-183445649 on Build GRCh38
rs138352332 Chr.4: 183445538-183445538 on Build GRCh38
rs1002760501 Chr.4: 183445584-183445584 on Build GRCh38
rs904423312 Chr.4: 183445487-183445487 on Build GRCh38
rs200915094 Chr.4: 183445478-183445478 on Build GRCh38
rs755155806 Chr.4: 183445542-183445542 on Build GRCh38
rs770954239 Chr.4: 183445654-183445654 on Build GRCh38
rs765236634 Chr.4: 183445526-183445526 on Build GRCh38
rs1025468818 Chr.4: 183445539-183445539 on Build GRCh38
rs764034598 Chr.4: 183445494-183445494 on Build GRCh38
rs908246633 Chr.4: 183445648-183445648 on Build GRCh38

View Full Product Details