Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23642840-23643107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN5
Entrez Gene ID
84516
Gene Name
dynactin subunit 5
Gene Aliases
-
UniGene
Hs.435941
Gene Chromosome Location
Chr.16: 23641366-23676207 on Build GRCh38
Transcripts
NM_001199011.1 NM_001199743.1 NM_032486.3 NR_037573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5631146 c.86G>C p.C29S Chr.16: 23642992-23642992 on Build GRCh38
3849548 c.56G>T p.G19V Chr.16: 23642962-23642962 on Build GRCh38
4940766 c.69T>C p.S23S Chr.16: 23642975-23642975 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373577774 Chr.16: 23642940-23642940 on Build GRCh38
rs369282000 Chr.16: 23642939-23642939 on Build GRCh38
rs776678809 Chr.16: 23642979-23642982 on Build GRCh38
rs747068474 Chr.16: 23643073-23643073 on Build GRCh38
rs528459331 Chr.16: 23643075-23643075 on Build GRCh38
rs141294424 Chr.16: 23642996-23642996 on Build GRCh38
rs762184764 Chr.16: 23642952-23642952 on Build GRCh38
rs375849466 Chr.16: 23642916-23642916 on Build GRCh38
rs117841286 Chr.16: 23642931-23642931 on Build GRCh38
rs755872761 Chr.16: 23643053-23643053 on Build GRCh38
rs376915818 Chr.16: 23642938-23642938 on Build GRCh38
rs750410754 Chr.16: 23642901-23642901 on Build GRCh38
rs201806949 Chr.16: 23642965-23642965 on Build GRCh38
rs768713201 Chr.16: 23642914-23642914 on Build GRCh38
rs56737329 Chr.16: 23643049-23643049 on Build GRCh38
rs765548172 Chr.16: 23642963-23642963 on Build GRCh38
rs371359620 Chr.16: 23642937-23642937 on Build GRCh38
rs751086676 Chr.16: 23643032-23643032 on Build GRCh38
rs747957368 Chr.16: 23642926-23642926 on Build GRCh38
rs546594210 Chr.16: 23643066-23643066 on Build GRCh38
rs1002897789 Chr.16: 23643046-23643046 on Build GRCh38
rs367830018 Chr.16: 23642934-23642934 on Build GRCh38
rs146108494 Chr.16: 23643051-23643051 on Build GRCh38
rs750022035 Chr.16: 23642968-23642968 on Build GRCh38
rs767873406 Chr.16: 23642891-23642891 on Build GRCh38
rs374046641 Chr.16: 23643074-23643074 on Build GRCh38
rs761687132 Chr.16: 23643070-23643071 on Build GRCh38
rs1020239196 Chr.16: 23642951-23642951 on Build GRCh38
rs920142421 Chr.16: 23643071-23643071 on Build GRCh38
rs142955912 Chr.16: 23642929-23642929 on Build GRCh38
rs772080456 Chr.16: 23642932-23642932 on Build GRCh38
rs749164288 Chr.16: 23643061-23643061 on Build GRCh38
rs769499535 Chr.16: 23642928-23642928 on Build GRCh38
rs996430480 Chr.16: 23642903-23642903 on Build GRCh38
rs746577412 Chr.16: 23642910-23642910 on Build GRCh38
rs772276721 Chr.16: 23643064-23643064 on Build GRCh38
rs754588165 Chr.16: 23643042-23643042 on Build GRCh38
rs768841757 Chr.16: 23643060-23643060 on Build GRCh38
rs768445164 Chr.16: 23642940-23642942 on Build GRCh38
rs779922842 Chr.16: 23643026-23643026 on Build GRCh38

View Full Product Details