Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76009417-76009672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVPL
Entrez Gene ID
2125
Gene Name
envoplakin
Gene Aliases
EVPK
UniGene
Hs.500635
Gene Chromosome Location
Chr.17: 76006845-76027452 on Build GRCh38
Transcripts
NM_001320747.1 NM_001988.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3820605 c.3743T>G p.V1248G Chr.17: 76009462-76009462 on Build GRCh38
6523906 c.3634C>T p.R1212W Chr.17: 76009571-76009571 on Build GRCh38
7366721 c.3727G>A p.A1243T Chr.17: 76009478-76009478 on Build GRCh38
6656958 c.3741G>A p.T1247T Chr.17: 76009464-76009464 on Build GRCh38
4757478 c.3721C>A p.L1241M Chr.17: 76009484-76009484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748460268 Chr.17: 76009615-76009615 on Build GRCh38
rs752000423 Chr.17: 76009573-76009573 on Build GRCh38
rs766924736 Chr.17: 76009571-76009571 on Build GRCh38
rs755721021 Chr.17: 76009581-76009581 on Build GRCh38
rs145713300 Chr.17: 76009465-76009465 on Build GRCh38
rs144952445 Chr.17: 76009622-76009622 on Build GRCh38
rs148100325 Chr.17: 76009480-76009480 on Build GRCh38
rs931441818 Chr.17: 76009528-76009528 on Build GRCh38
rs770762901 Chr.17: 76009522-76009522 on Build GRCh38
rs752994504 Chr.17: 76009645-76009645 on Build GRCh38
rs907576435 Chr.17: 76009459-76009459 on Build GRCh38
rs367555955 Chr.17: 76009542-76009542 on Build GRCh38
rs370565249 Chr.17: 76009593-76009593 on Build GRCh38
rs148648557 Chr.17: 76009640-76009640 on Build GRCh38
rs751160453 Chr.17: 76009603-76009603 on Build GRCh38
rs777119774 Chr.17: 76009520-76009520 on Build GRCh38
rs139861901 Chr.17: 76009550-76009550 on Build GRCh38
rs759928902 Chr.17: 76009574-76009574 on Build GRCh38
rs554283625 Chr.17: 76009478-76009478 on Build GRCh38
rs749319043 Chr.17: 76009521-76009521 on Build GRCh38
rs763527200 Chr.17: 76009570-76009570 on Build GRCh38
rs200731297 Chr.17: 76009529-76009529 on Build GRCh38
rs770804170 Chr.17: 76009453-76009453 on Build GRCh38
rs778783709 Chr.17: 76009600-76009600 on Build GRCh38
rs762898947 Chr.17: 76009621-76009621 on Build GRCh38
rs773149269 Chr.17: 76009440-76009440 on Build GRCh38
rs920183082 Chr.17: 76009499-76009499 on Build GRCh38
rs755695873 Chr.17: 76009498-76009498 on Build GRCh38
rs769982483 Chr.17: 76009544-76009544 on Build GRCh38
rs559827414 Chr.17: 76009629-76009629 on Build GRCh38
rs774132608 Chr.17: 76009460-76009460 on Build GRCh38
rs147527679 Chr.17: 76009586-76009586 on Build GRCh38
rs139479728 Chr.17: 76009604-76009604 on Build GRCh38
rs970400848 Chr.17: 76009583-76009583 on Build GRCh38
rs767967549 Chr.17: 76009575-76009575 on Build GRCh38
rs780949433 Chr.17: 76009497-76009497 on Build GRCh38
rs749155048 Chr.17: 76009450-76009450 on Build GRCh38
rs201966558 Chr.17: 76009614-76009614 on Build GRCh38
rs866389061 Chr.17: 76009482-76009482 on Build GRCh38
rs375245505 Chr.17: 76009620-76009620 on Build GRCh38
rs370996950 Chr.17: 76009468-76009468 on Build GRCh38
rs745685608 Chr.17: 76009607-76009607 on Build GRCh38
rs375552944 Chr.17: 76009496-76009496 on Build GRCh38
rs141948489 Chr.17: 76009481-76009481 on Build GRCh38
rs144978675 Chr.17: 76009619-76009619 on Build GRCh38
rs534647471 Chr.17: 76009464-76009464 on Build GRCh38
rs75019874 Chr.17: 76009530-76009530 on Build GRCh38
rs368923796 Chr.17: 76009613-76009613 on Build GRCh38
rs200046392 Chr.17: 76009541-76009541 on Build GRCh38
rs761919888 Chr.17: 76009543-76009543 on Build GRCh38
rs768065645 Chr.17: 76009644-76009644 on Build GRCh38
rs143353839 Chr.17: 76009580-76009580 on Build GRCh38
rs376792152 Chr.17: 76009561-76009561 on Build GRCh38
rs1014644565 Chr.17: 76009649-76009649 on Build GRCh38
rs867136088 Chr.17: 76009451-76009451 on Build GRCh38
rs747762061 Chr.17: 76009573-76009573 on Build GRCh38
rs776697613 Chr.17: 76009467-76009467 on Build GRCh38

View Full Product Details