Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 108364517-108364790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL17
Entrez Gene ID
64839
Gene Name
F-box and leucine rich repeat protein 17
Gene Aliases
FBXO13, Fbl17, Fbx13
UniGene
Hs.657225
Gene Chromosome Location
Chr.5: 107859033-108382098 on Build GRCh38
Transcripts
NM_001163315.2 XM_005272048.4 XM_005272050.4 XM_011543574.2 XM_011543575.2 XM_011543576.2 XM_011543577.2 XM_011543578.2 XM_011543579.2 XM_011543580.2 XM_017009729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5665243 c.1339A>C p.N447H Chr.5: 108364773-108364773 on Build GRCh38
5665245 c.145A>C p.N49H Chr.5: 108364773-108364773 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs957422851 Chr.5: 108364683-108364683 on Build GRCh38
rs756230874 Chr.5: 108364725-108364725 on Build GRCh38
rs751404654 Chr.5: 108364721-108364721 on Build GRCh38
rs890084307 Chr.5: 108364544-108364544 on Build GRCh38
rs1026540589 Chr.5: 108364584-108364584 on Build GRCh38
rs754270388 Chr.5: 108364590-108364590 on Build GRCh38
rs112875633 Chr.5: 108364756-108364756 on Build GRCh38
rs755015367 Chr.5: 108364729-108364729 on Build GRCh38
rs970028126 Chr.5: 108364644-108364644 on Build GRCh38
rs755374008 Chr.5: 108364612-108364612 on Build GRCh38
rs1024342070 Chr.5: 108364639-108364639 on Build GRCh38
rs561860895 Chr.5: 108364668-108364668 on Build GRCh38
rs758934458 Chr.5: 108364744-108364744 on Build GRCh38
rs371972809 Chr.5: 108364718-108364718 on Build GRCh38
rs185672068 Chr.5: 108364581-108364581 on Build GRCh38
rs1029603818 Chr.5: 108364693-108364693 on Build GRCh38
rs1014552201 Chr.5: 108364606-108364606 on Build GRCh38
rs1007351834 Chr.5: 108364546-108364546 on Build GRCh38
rs189005113 Chr.5: 108364677-108364677 on Build GRCh38
rs748603942 Chr.5: 108364742-108364742 on Build GRCh38
rs980178423 Chr.5: 108364647-108364647 on Build GRCh38
rs865784316 Chr.5: 108364658-108364658 on Build GRCh38
rs1056540814 Chr.5: 108364552-108364552 on Build GRCh38
rs779012594 Chr.5: 108364741-108364741 on Build GRCh38
rs547192092 Chr.5: 108364684-108364684 on Build GRCh38
rs541829911 Chr.5: 108364602-108364602 on Build GRCh38
rs985318588 Chr.5: 108364727-108364727 on Build GRCh38
rs531707531 Chr.5: 108364595-108364595 on Build GRCh38
rs764908227 Chr.5: 108364705-108364705 on Build GRCh38
rs775538364 Chr.5: 108364711-108364711 on Build GRCh38
rs899024098 Chr.5: 108364571-108364571 on Build GRCh38
rs570311491 Chr.5: 108364716-108364716 on Build GRCh38
rs756707081 Chr.5: 108364558-108364558 on Build GRCh38
rs1032709667 Chr.5: 108364650-108364651 on Build GRCh38
rs948261307 Chr.5: 108364559-108364559 on Build GRCh38
rs1010180855 Chr.5: 108364659-108364659 on Build GRCh38
rs766608138 Chr.5: 108364726-108364726 on Build GRCh38
rs759074259 Chr.5: 108364700-108364700 on Build GRCh38

View Full Product Details