Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147758774-147759009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA5
Entrez Gene ID
2702
Gene Name
gap junction protein alpha 5
Gene Aliases
ATFB11, CX40
UniGene
Hs.447968
Gene Chromosome Location
Chr.1: 147756199-147781127 on Build GRCh38
Transcripts
NM_005266.6 NM_181703.3 XM_005272951.4 XM_017001044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
395111 c.353G>A p.R118Q Chr.1: 147758886-147758886 on Build GRCh38
1127404 c.352C>T p.R118W Chr.1: 147758887-147758887 on Build GRCh38
6500672 c.281G>C p.G94A Chr.1: 147758958-147758958 on Build GRCh38
895327 c.327G>T p.R109R Chr.1: 147758912-147758912 on Build GRCh38
895328 c.296C>A p.T99N Chr.1: 147758943-147758943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782281038 Chr.1: 147758978-147758978 on Build GRCh38
rs782334563 Chr.1: 147758908-147758908 on Build GRCh38
rs121434558 Chr.1: 147758977-147758977 on Build GRCh38
rs782394288 Chr.1: 147758984-147758984 on Build GRCh38
rs782780433 Chr.1: 147758862-147758862 on Build GRCh38
rs781802553 Chr.1: 147758806-147758806 on Build GRCh38
rs782770843 Chr.1: 147758840-147758840 on Build GRCh38
rs781832760 Chr.1: 147758971-147758971 on Build GRCh38
rs886045251 Chr.1: 147758897-147758897 on Build GRCh38
rs1018708274 Chr.1: 147758967-147758967 on Build GRCh38
rs782415947 Chr.1: 147758981-147758981 on Build GRCh38
rs781866205 Chr.1: 147758950-147758950 on Build GRCh38
rs782301284 Chr.1: 147758937-147758937 on Build GRCh38
rs886045252 Chr.1: 147758905-147758905 on Build GRCh38
rs782546377 Chr.1: 147758892-147758892 on Build GRCh38
rs782435078 Chr.1: 147758966-147758966 on Build GRCh38
rs782438073 Chr.1: 147758809-147758809 on Build GRCh38
rs965871957 Chr.1: 147758990-147758990 on Build GRCh38
rs375921833 Chr.1: 147758951-147758951 on Build GRCh38
rs782247905 Chr.1: 147758913-147758913 on Build GRCh38
rs782509380 Chr.1: 147758975-147758975 on Build GRCh38
rs781905640 Chr.1: 147758959-147758959 on Build GRCh38
rs782181477 Chr.1: 147758906-147758906 on Build GRCh38
rs374342167 Chr.1: 147758964-147758964 on Build GRCh38
rs138375318 Chr.1: 147758940-147758940 on Build GRCh38
rs150168016 Chr.1: 147758891-147758891 on Build GRCh38
rs587665554 Chr.1: 147758968-147758968 on Build GRCh38
rs201082105 Chr.1: 147758842-147758842 on Build GRCh38
rs782013554 Chr.1: 147758915-147758915 on Build GRCh38
rs782254259 Chr.1: 147758942-147758943 on Build GRCh38
rs112980895 Chr.1: 147758861-147758861 on Build GRCh38
rs1010609800 Chr.1: 147758930-147758930 on Build GRCh38
rs782392307 Chr.1: 147758914-147758914 on Build GRCh38
rs782273824 Chr.1: 147758796-147758796 on Build GRCh38
rs782675678 Chr.1: 147758904-147758904 on Build GRCh38
rs782082372 Chr.1: 147758824-147758824 on Build GRCh38
rs121434557 Chr.1: 147758953-147758953 on Build GRCh38
rs976822450 Chr.1: 147758823-147758823 on Build GRCh38
rs375331306 Chr.1: 147758900-147758900 on Build GRCh38
rs906714543 Chr.1: 147758888-147758888 on Build GRCh38
rs139275267 Chr.1: 147758868-147758868 on Build GRCh38
rs2232191 Chr.1: 147758870-147758870 on Build GRCh38
rs782428701 Chr.1: 147758869-147758869 on Build GRCh38
rs782580208 Chr.1: 147758886-147758886 on Build GRCh38
rs143695986 Chr.1: 147758912-147758912 on Build GRCh38
rs781923468 Chr.1: 147758814-147758814 on Build GRCh38
rs781987662 Chr.1: 147758847-147758847 on Build GRCh38
rs782642890 Chr.1: 147758881-147758881 on Build GRCh38
rs782730854 Chr.1: 147758826-147758826 on Build GRCh38
rs1045225638 Chr.1: 147758882-147758882 on Build GRCh38
rs387906613 Chr.1: 147758986-147758986 on Build GRCh38
rs782647757 Chr.1: 147758976-147758976 on Build GRCh38
rs367924086 Chr.1: 147758922-147758922 on Build GRCh38
rs1003271827 Chr.1: 147758889-147758889 on Build GRCh38
rs370874304 Chr.1: 147758841-147758841 on Build GRCh38

View Full Product Details