Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73443115-73443382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REC114
Entrez Gene ID
283677
Gene Name
REC114 meiotic recombination protein
Gene Aliases
C15orf60, CT147
UniGene
Hs.743574
Gene Chromosome Location
Chr.15: 73443158-73560015 on Build GRCh38
Transcripts
NM_001042367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1735276 c.147C>A p.C49* Chr.15: 73443332-73443332 on Build GRCh38
7358092 c.104G>T p.R35I Chr.15: 73443289-73443289 on Build GRCh38
3988117 c.126G>T p.L42L Chr.15: 73443311-73443311 on Build GRCh38
1317764 c.4G>A p.A2T Chr.15: 73443189-73443189 on Build GRCh38
3744742 c.62G>A p.W21* Chr.15: 73443247-73443247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757153075 Chr.15: 73443349-73443349 on Build GRCh38
rs767801783 Chr.15: 73443158-73443158 on Build GRCh38
rs775202410 Chr.15: 73443208-73443208 on Build GRCh38
rs768441032 Chr.15: 73443210-73443210 on Build GRCh38
rs992289597 Chr.15: 73443184-73443184 on Build GRCh38
rs779792470 Chr.15: 73443268-73443268 on Build GRCh38
rs769623663 Chr.15: 73443206-73443206 on Build GRCh38
rs751627032 Chr.15: 73443154-73443154 on Build GRCh38
rs762018713 Chr.15: 73443157-73443157 on Build GRCh38
rs1026068674 Chr.15: 73443280-73443280 on Build GRCh38
rs774454922 Chr.15: 73443214-73443214 on Build GRCh38
rs148736034 Chr.15: 73443170-73443170 on Build GRCh38
rs767746609 Chr.15: 73443222-73443222 on Build GRCh38
rs903501042 Chr.15: 73443237-73443237 on Build GRCh38
rs371048409 Chr.15: 73443250-73443250 on Build GRCh38
rs756371614 Chr.15: 73443239-73443239 on Build GRCh38
rs377562089 Chr.15: 73443243-73443243 on Build GRCh38
rs962845077 Chr.15: 73443160-73443160 on Build GRCh38
rs914165545 Chr.15: 73443203-73443203 on Build GRCh38
rs764217700 Chr.15: 73443153-73443153 on Build GRCh38
rs745556964 Chr.15: 73443204-73443204 on Build GRCh38
rs745516975 Chr.15: 73443264-73443264 on Build GRCh38
rs754084628 Chr.15: 73443242-73443242 on Build GRCh38
rs779342064 Chr.15: 73443247-73443247 on Build GRCh38
rs72741460 Chr.15: 73443182-73443182 on Build GRCh38
rs113268207 Chr.15: 73443354-73443354 on Build GRCh38
rs779040927 Chr.15: 73443348-73443348 on Build GRCh38
rs535083559 Chr.15: 73443168-73443168 on Build GRCh38
rs867090516 Chr.15: 73443295-73443295 on Build GRCh38
rs1016959027 Chr.15: 73443338-73443338 on Build GRCh38
rs575946798 Chr.15: 73443233-73443233 on Build GRCh38
rs576871413 Chr.15: 73443305-73443305 on Build GRCh38
rs370941071 Chr.15: 73443218-73443218 on Build GRCh38
rs748165364 Chr.15: 73443299-73443299 on Build GRCh38
rs757491816 Chr.15: 73443340-73443340 on Build GRCh38
rs776934489 Chr.15: 73443331-73443331 on Build GRCh38
rs540205218 Chr.15: 73443227-73443227 on Build GRCh38
rs541072358 Chr.15: 73443319-73443319 on Build GRCh38
rs780351209 Chr.15: 73443166-73443166 on Build GRCh38
rs978856610 Chr.15: 73443164-73443164 on Build GRCh38
rs753698315 Chr.15: 73443345-73443345 on Build GRCh38
rs1006527585 Chr.15: 73443330-73443330 on Build GRCh38
rs768625072 Chr.15: 73443281-73443281 on Build GRCh38
rs759796022 Chr.15: 73443332-73443332 on Build GRCh38
rs761965215 Chr.15: 73443219-73443219 on Build GRCh38
rs370726487 Chr.15: 73443136-73443136 on Build GRCh38
rs762814881 Chr.15: 73443209-73443209 on Build GRCh38
rs187528283 Chr.15: 73443137-73443137 on Build GRCh38
rs748990746 Chr.15: 73443273-73443273 on Build GRCh38
rs780798070 Chr.15: 73443190-73443190 on Build GRCh38
rs926049036 Chr.15: 73443288-73443288 on Build GRCh38
rs570878291 Chr.15: 73443143-73443143 on Build GRCh38
rs760814616 Chr.15: 73443315-73443315 on Build GRCh38
rs950322860 Chr.15: 73443186-73443186 on Build GRCh38
rs374263335 Chr.15: 73443240-73443240 on Build GRCh38
rs762862189 Chr.15: 73443148-73443148 on Build GRCh38
rs773369299 Chr.15: 73443310-73443310 on Build GRCh38
rs780744581 Chr.15: 73443253-73443253 on Build GRCh38
rs769268662 Chr.15: 73443267-73443267 on Build GRCh38
rs750671215 Chr.15: 73443226-73443226 on Build GRCh38
rs753250701 Chr.15: 73443249-73443249 on Build GRCh38
rs765628791 Chr.15: 73443343-73443343 on Build GRCh38
rs909844684 Chr.15: 73443150-73443150 on Build GRCh38
rs574730813 Chr.15: 73443178-73443178 on Build GRCh38
rs764595003 Chr.15: 73443350-73443350 on Build GRCh38
rs552281945 Chr.15: 73443139-73443139 on Build GRCh38
rs749865235 Chr.15: 73443353-73443353 on Build GRCh38
rs774401829 Chr.15: 73443282-73443282 on Build GRCh38
rs755383502 Chr.15: 73443169-73443169 on Build GRCh38

View Full Product Details