Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159066001-159066270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIM2
Entrez Gene ID
9447
Gene Name
absent in melanoma 2
Gene Aliases
PYHIN4
UniGene
Hs.733411
Gene Chromosome Location
Chr.1: 159059226-159132351 on Build GRCh38
Transcripts
NM_004833.1 XM_005245616.4 XM_017002848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
676000 c.564C>T p.F188F Chr.1: 159066162-159066162 on Build GRCh38
318612 c.548C>A p.T183K Chr.1: 159066178-159066178 on Build GRCh38
4170561 c.521A>C p.Q174P Chr.1: 159066205-159066205 on Build GRCh38
278726 c.550G>T p.E184* Chr.1: 159066176-159066176 on Build GRCh38
5987770 c.700C>T p.Q234* Chr.1: 159066026-159066026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761571921 Chr.1: 159066164-159066164 on Build GRCh38
rs760089097 Chr.1: 159066145-159066145 on Build GRCh38
rs770609652 Chr.1: 159066103-159066103 on Build GRCh38
rs373555103 Chr.1: 159066243-159066243 on Build GRCh38
rs756118457 Chr.1: 159066193-159066193 on Build GRCh38
rs376898750 Chr.1: 159066035-159066035 on Build GRCh38
rs35130877 Chr.1: 159066168-159066168 on Build GRCh38
rs756277653 Chr.1: 159066054-159066054 on Build GRCh38
rs772001101 Chr.1: 159066141-159066141 on Build GRCh38
rs572476207 Chr.1: 159066207-159066207 on Build GRCh38
rs148686373 Chr.1: 159066057-159066057 on Build GRCh38
rs969176404 Chr.1: 159066182-159066182 on Build GRCh38
rs368263314 Chr.1: 159066175-159066175 on Build GRCh38
rs775282472 Chr.1: 159066239-159066239 on Build GRCh38
rs866050462 Chr.1: 159066220-159066220 on Build GRCh38
rs375624593 Chr.1: 159066048-159066048 on Build GRCh38
rs774155198 Chr.1: 159066112-159066112 on Build GRCh38
rs375608638 Chr.1: 159066181-159066181 on Build GRCh38
rs763723491 Chr.1: 159066146-159066146 on Build GRCh38
rs745765084 Chr.1: 159066138-159066138 on Build GRCh38
rs774921419 Chr.1: 159066143-159066143 on Build GRCh38
rs545837561 Chr.1: 159066228-159066228 on Build GRCh38
rs539742230 Chr.1: 159066069-159066069 on Build GRCh38
rs771911036 Chr.1: 159066230-159066230 on Build GRCh38
rs369765206 Chr.1: 159066049-159066049 on Build GRCh38
rs944682755 Chr.1: 159066064-159066064 on Build GRCh38
rs745673444 Chr.1: 159066229-159066229 on Build GRCh38
rs200913901 Chr.1: 159066171-159066171 on Build GRCh38
rs765563535 Chr.1: 159066089-159066090 on Build GRCh38
rs749567312 Chr.1: 159066085-159066085 on Build GRCh38
rs1013616810 Chr.1: 159066159-159066159 on Build GRCh38
rs757503647 Chr.1: 159066216-159066216 on Build GRCh38
rs1052245685 Chr.1: 159066245-159066245 on Build GRCh38
rs768127211 Chr.1: 159066178-159066178 on Build GRCh38
rs373307771 Chr.1: 159066047-159066047 on Build GRCh38
rs748193319 Chr.1: 159066050-159066050 on Build GRCh38
rs776114459 Chr.1: 159066156-159066156 on Build GRCh38

View Full Product Details