Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6447682-6447951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM64A
Entrez Gene ID
54478
Gene Name
family with sequence similarity 64 member A
Gene Aliases
CATS, RCS1
UniGene
Hs.592116
Gene Chromosome Location
Chr.17: 6444305-6451065 on Build GRCh38
Transcripts
NM_001195228.1 NM_019013.2 XM_011523944.2 XM_017024778.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4683123 c.526C>T p.R176* Chr.17: 6447694-6447694 on Build GRCh38
4683124 c.526C>T p.R176* Chr.17: 6447694-6447694 on Build GRCh38
983019 c.524G>A p.R175Q Chr.17: 6447692-6447692 on Build GRCh38
5029607 c.556C>G p.P186A Chr.17: 6447724-6447724 on Build GRCh38
5029608 c.556C>G p.P186A Chr.17: 6447724-6447724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35213363 Chr.17: 6447726-6447726 on Build GRCh38
rs772336733 Chr.17: 6447802-6447802 on Build GRCh38
rs749827421 Chr.17: 6447712-6447712 on Build GRCh38
rs756728002 Chr.17: 6447799-6447799 on Build GRCh38
rs909693352 Chr.17: 6447803-6447803 on Build GRCh38
rs767288905 Chr.17: 6447773-6447773 on Build GRCh38
rs16955866 Chr.17: 6447801-6447801 on Build GRCh38
rs947186048 Chr.17: 6447752-6447752 on Build GRCh38
rs751512933 Chr.17: 6447760-6447760 on Build GRCh38
rs754790121 Chr.17: 6447769-6447769 on Build GRCh38
rs763003347 Chr.17: 6447750-6447750 on Build GRCh38
rs774901405 Chr.17: 6447719-6447719 on Build GRCh38
rs368256466 Chr.17: 6447793-6447793 on Build GRCh38
rs565789654 Chr.17: 6447906-6447906 on Build GRCh38
rs776808945 Chr.17: 6447747-6447747 on Build GRCh38
rs61457133 Chr.17: 6447815-6447815 on Build GRCh38
rs753396677 Chr.17: 6447796-6447796 on Build GRCh38
rs764270316 Chr.17: 6447742-6447742 on Build GRCh38
rs946965009 Chr.17: 6447916-6447916 on Build GRCh38
rs752355469 Chr.17: 6447782-6447782 on Build GRCh38
rs898972807 Chr.17: 6447877-6447877 on Build GRCh38
rs771379702 Chr.17: 6447718-6447718 on Build GRCh38
rs368127473 Chr.17: 6447733-6447733 on Build GRCh38
rs933639182 Chr.17: 6447895-6447895 on Build GRCh38
rs767139439 Chr.17: 6447914-6447915 on Build GRCh38
rs533450254 Chr.17: 6447756-6447756 on Build GRCh38
rs564586671 Chr.17: 6447755-6447755 on Build GRCh38
rs200670174 Chr.17: 6447703-6447703 on Build GRCh38
rs771279948 Chr.17: 6447840-6447840 on Build GRCh38
rs995641504 Chr.17: 6447905-6447905 on Build GRCh38
rs902890418 Chr.17: 6447785-6447785 on Build GRCh38
rs188898430 Chr.17: 6447851-6447851 on Build GRCh38
rs780394827 Chr.17: 6447808-6447808 on Build GRCh38
rs760922295 Chr.17: 6447741-6447741 on Build GRCh38
rs775632055 Chr.17: 6447739-6447739 on Build GRCh38
rs199975875 Chr.17: 6447783-6447783 on Build GRCh38
rs527690413 Chr.17: 6447704-6447704 on Build GRCh38
rs951462545 Chr.17: 6447907-6447907 on Build GRCh38

View Full Product Details