Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 100061151-100061396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN5
Entrez Gene ID
53942
Gene Name
contactin 5
Gene Aliases
HNB-2s, NB-2
UniGene
Hs.656783
Gene Chromosome Location
Chr.11: 99020904-100358885 on Build GRCh38
Transcripts
NM_001243270.1 NM_001243271.1 NM_014361.3 NM_175566.2 XM_011542871.1 XM_011542873.1 XM_017017926.1 XM_017017927.1 XM_017017928.1 XM_017017929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4629859 c.1108A>G p.R370G Chr.11: 100061339-100061339 on Build GRCh38
6070723 c.1142G>T p.R381L Chr.11: 100061373-100061373 on Build GRCh38
6631812 c.1056G>A p.A352A Chr.11: 100061287-100061287 on Build GRCh38
5684021 c.1063G>T p.E355* Chr.11: 100061294-100061294 on Build GRCh38
3454902 c.1127G>A p.G376E Chr.11: 100061358-100061358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772467575 Chr.11: 100061239-100061239 on Build GRCh38
rs557679651 Chr.11: 100061340-100061340 on Build GRCh38
rs201520977 Chr.11: 100061333-100061333 on Build GRCh38
rs201801323 Chr.11: 100061222-100061222 on Build GRCh38
rs201910584 Chr.11: 100061213-100061213 on Build GRCh38
rs769061773 Chr.11: 100061302-100061302 on Build GRCh38
rs772530310 Chr.11: 100061193-100061193 on Build GRCh38
rs762294192 Chr.11: 100061247-100061247 on Build GRCh38
rs200368105 Chr.11: 100061237-100061237 on Build GRCh38
rs775890074 Chr.11: 100061240-100061240 on Build GRCh38
rs374384868 Chr.11: 100061208-100061208 on Build GRCh38
rs201982881 Chr.11: 100061356-100061356 on Build GRCh38
rs779108160 Chr.11: 100061287-100061287 on Build GRCh38
rs200224105 Chr.11: 100061291-100061291 on Build GRCh38
rs866845091 Chr.11: 100061367-100061367 on Build GRCh38
rs199997328 Chr.11: 100061205-100061205 on Build GRCh38
rs199969666 Chr.11: 100061215-100061215 on Build GRCh38
rs369348604 Chr.11: 100061257-100061257 on Build GRCh38
rs113132209 Chr.11: 100061207-100061207 on Build GRCh38
rs186615197 Chr.11: 100061278-100061278 on Build GRCh38
rs200717963 Chr.11: 100061355-100061355 on Build GRCh38
rs755878727 Chr.11: 100061273-100061273 on Build GRCh38
rs777432657 Chr.11: 100061274-100061274 on Build GRCh38
rs201683115 Chr.11: 100061263-100061263 on Build GRCh38
rs200797524 Chr.11: 100061322-100061322 on Build GRCh38
rs904002387 Chr.11: 100061283-100061283 on Build GRCh38
rs767026275 Chr.11: 100061354-100061354 on Build GRCh38
rs376839752 Chr.11: 100061256-100061256 on Build GRCh38
rs200403626 Chr.11: 100061268-100061268 on Build GRCh38
rs754338921 Chr.11: 100061198-100061198 on Build GRCh38
rs754683207 Chr.11: 100061183-100061183 on Build GRCh38
rs56122118 Chr.11: 100061301-100061301 on Build GRCh38
rs745905925 Chr.11: 100061288-100061288 on Build GRCh38
rs775800095 Chr.11: 100061293-100061293 on Build GRCh38
rs796084913 Chr.11: 100061188-100061188 on Build GRCh38
rs199796384 Chr.11: 100061286-100061286 on Build GRCh38
rs754593231 Chr.11: 100061258-100061258 on Build GRCh38
rs199552993 Chr.11: 100061339-100061339 on Build GRCh38
rs267603261 Chr.11: 100061281-100061281 on Build GRCh38
rs199698378 Chr.11: 100061306-100061306 on Build GRCh38
rs765773786 Chr.11: 100061323-100061323 on Build GRCh38
rs200767659 Chr.11: 100061246-100061246 on Build GRCh38
rs1012235370 Chr.11: 100061276-100061276 on Build GRCh38
rs182344400 Chr.11: 100061216-100061216 on Build GRCh38
rs776919295 Chr.11: 100061305-100061305 on Build GRCh38

View Full Product Details