Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53305871-53306144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf10
Entrez Gene ID
60314
Gene Name
chromosome 12 open reading frame 10
Gene Aliases
Gamm1, MST024, MSTP024, MYG, MYG1
UniGene
Hs.655988
Gene Chromosome Location
Chr.12: 53299686-53307181 on Build GRCh38
Transcripts
NM_021640.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1197945 c.563G>A p.R188Q Chr.12: 53305981-53305981 on Build GRCh38
3812370 c.623A>C p.H208P Chr.12: 53306041-53306041 on Build GRCh38
7155957 c.640G>C p.E214Q Chr.12: 53306058-53306058 on Build GRCh38
1932861 c.592C>T p.R198* Chr.12: 53306010-53306010 on Build GRCh38
140691 c.562C>T p.R188* Chr.12: 53305980-53305980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776717369 Chr.12: 53305908-53305908 on Build GRCh38
rs1041294225 Chr.12: 53305895-53305895 on Build GRCh38
rs936484063 Chr.12: 53306093-53306093 on Build GRCh38
rs768179519 Chr.12: 53306082-53306082 on Build GRCh38
rs770019093 Chr.12: 53305929-53305929 on Build GRCh38
rs200946706 Chr.12: 53306064-53306064 on Build GRCh38
rs975010686 Chr.12: 53306116-53306116 on Build GRCh38
rs148056462 Chr.12: 53306011-53306011 on Build GRCh38
rs1046369630 Chr.12: 53306022-53306022 on Build GRCh38
rs760701836 Chr.12: 53306113-53306113 on Build GRCh38
rs373655066 Chr.12: 53306091-53306091 on Build GRCh38
rs376890469 Chr.12: 53306082-53306082 on Build GRCh38
rs761747121 Chr.12: 53306107-53306108 on Build GRCh38
rs774077592 Chr.12: 53306007-53306007 on Build GRCh38
rs778731948 Chr.12: 53306043-53306043 on Build GRCh38
rs147017503 Chr.12: 53305994-53305994 on Build GRCh38
rs775772521 Chr.12: 53306002-53306002 on Build GRCh38
rs539662599 Chr.12: 53306001-53306001 on Build GRCh38
rs931059921 Chr.12: 53305924-53305924 on Build GRCh38
rs780245809 Chr.12: 53305974-53305974 on Build GRCh38
rs374242491 Chr.12: 53305922-53305922 on Build GRCh38
rs897428002 Chr.12: 53305905-53305905 on Build GRCh38
rs763938406 Chr.12: 53305940-53305940 on Build GRCh38
rs200623152 Chr.12: 53305997-53305997 on Build GRCh38
rs746484118 Chr.12: 53305942-53305943 on Build GRCh38
rs776496605 Chr.12: 53306108-53306108 on Build GRCh38
rs374431799 Chr.12: 53306020-53306020 on Build GRCh38
rs750371967 Chr.12: 53306016-53306016 on Build GRCh38
rs528978516 Chr.12: 53305952-53305952 on Build GRCh38
rs768873299 Chr.12: 53306006-53306006 on Build GRCh38
rs775615604 Chr.12: 53305932-53305932 on Build GRCh38
rs747388893 Chr.12: 53305980-53305980 on Build GRCh38
rs753491194 Chr.12: 53305972-53305972 on Build GRCh38
rs567684856 Chr.12: 53305910-53305910 on Build GRCh38
rs142971160 Chr.12: 53305981-53305981 on Build GRCh38
rs761624531 Chr.12: 53306010-53306010 on Build GRCh38
rs907199326 Chr.12: 53306045-53306045 on Build GRCh38
rs1019861018 Chr.12: 53306111-53306111 on Build GRCh38
rs368245768 Chr.12: 53305949-53305949 on Build GRCh38
rs376935988 Chr.12: 53306100-53306100 on Build GRCh38
rs762717810 Chr.12: 53305934-53305934 on Build GRCh38
rs201601756 Chr.12: 53306066-53306066 on Build GRCh38
rs1027085763 Chr.12: 53306118-53306118 on Build GRCh38
rs779729576 Chr.12: 53305965-53305965 on Build GRCh38
rs1027188412 Chr.12: 53305942-53305942 on Build GRCh38
rs371992654 Chr.12: 53306019-53306019 on Build GRCh38
rs112202958 Chr.12: 53306084-53306084 on Build GRCh38
rs371378902 Chr.12: 53306032-53306032 on Build GRCh38
rs937356878 Chr.12: 53306046-53306046 on Build GRCh38
rs773196162 Chr.12: 53306109-53306109 on Build GRCh38
rs763232467 Chr.12: 53306005-53306005 on Build GRCh38
rs754618796 Chr.12: 53306033-53306033 on Build GRCh38
rs755617027 Chr.12: 53305961-53305961 on Build GRCh38
rs756218290 Chr.12: 53306068-53306068 on Build GRCh38
rs566779163 Chr.12: 53306108-53306108 on Build GRCh38
rs771567139 Chr.12: 53305897-53305897 on Build GRCh38
rs778214873 Chr.12: 53305978-53305978 on Build GRCh38
rs370034772 Chr.12: 53306054-53306054 on Build GRCh38
rs749905402 Chr.12: 53305955-53305955 on Build GRCh38
rs375977138 Chr.12: 53306048-53306048 on Build GRCh38
rs372971018 Chr.12: 53306085-53306085 on Build GRCh38

View Full Product Details