Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94260064-94260337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEM
Entrez Gene ID
2669
Gene Name
GTP binding protein overexpressed in skeletal muscle
Gene Aliases
KIR
UniGene
Hs.654463
Gene Chromosome Location
Chr.8: 94249249-94262345 on Build GRCh38
Transcripts
NM_005261.3 NM_181702.2 XM_017013315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
296578 c.167G>A p.R56Q Chr.8: 94260337-94260337 on Build GRCh38
752483 c.271C>T p.L91L Chr.8: 94260233-94260233 on Build GRCh38
5587865 c.203C>T p.S68F Chr.8: 94260301-94260301 on Build GRCh38
3651913 c.254G>A p.G85E Chr.8: 94260250-94260250 on Build GRCh38
3651912 c.274G>A p.A92T Chr.8: 94260230-94260230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139494082 Chr.8: 94260251-94260251 on Build GRCh38
rs887416077 Chr.8: 94260227-94260227 on Build GRCh38
rs766109484 Chr.8: 94260285-94260285 on Build GRCh38
rs763246450 Chr.8: 94260197-94260201 on Build GRCh38
rs1047451181 Chr.8: 94260211-94260211 on Build GRCh38
rs747460145 Chr.8: 94260267-94260267 on Build GRCh38
rs768860314 Chr.8: 94260269-94260269 on Build GRCh38
rs558816237 Chr.8: 94260172-94260172 on Build GRCh38
rs764970254 Chr.8: 94260188-94260188 on Build GRCh38
rs376694011 Chr.8: 94260148-94260148 on Build GRCh38
rs550393204 Chr.8: 94260275-94260275 on Build GRCh38
rs762025958 Chr.8: 94260187-94260187 on Build GRCh38
rs780616166 Chr.8: 94260234-94260234 on Build GRCh38
rs780005500 Chr.8: 94260194-94260194 on Build GRCh38
rs751132195 Chr.8: 94260199-94260199 on Build GRCh38
rs772521690 Chr.8: 94260255-94260255 on Build GRCh38
rs747744765 Chr.8: 94260127-94260127 on Build GRCh38
rs761960990 Chr.8: 94260274-94260274 on Build GRCh38
rs747832501 Chr.8: 94260236-94260236 on Build GRCh38
rs780300506 Chr.8: 94260088-94260089 on Build GRCh38
rs774740012 Chr.8: 94260147-94260147 on Build GRCh38
rs772917669 Chr.8: 94260280-94260280 on Build GRCh38
rs776810929 Chr.8: 94260270-94260270 on Build GRCh38
rs780164919 Chr.8: 94260300-94260300 on Build GRCh38
rs751506282 Chr.8: 94260289-94260289 on Build GRCh38
rs201303048 Chr.8: 94260253-94260253 on Build GRCh38
rs916486616 Chr.8: 94260248-94260248 on Build GRCh38
rs946600589 Chr.8: 94260276-94260276 on Build GRCh38
rs758462055 Chr.8: 94260140-94260140 on Build GRCh38
rs1028214537 Chr.8: 94260099-94260099 on Build GRCh38
rs759984927 Chr.8: 94260150-94260150 on Build GRCh38
rs762800000 Chr.8: 94260283-94260283 on Build GRCh38
rs950917586 Chr.8: 94260109-94260109 on Build GRCh38
rs973693867 Chr.8: 94260098-94260098 on Build GRCh38
rs767038807 Chr.8: 94260307-94260307 on Build GRCh38
rs754873278 Chr.8: 94260298-94260298 on Build GRCh38
rs377348768 Chr.8: 94260154-94260154 on Build GRCh38
rs536135668 Chr.8: 94260094-94260094 on Build GRCh38
rs913888309 Chr.8: 94260133-94260133 on Build GRCh38
rs754124324 Chr.8: 94260186-94260186 on Build GRCh38
rs765472126 Chr.8: 94260278-94260278 on Build GRCh38
rs754500839 Chr.8: 94260208-94260208 on Build GRCh38
rs776127220 Chr.8: 94260259-94260259 on Build GRCh38
rs567967510 Chr.8: 94260250-94260250 on Build GRCh38
rs771206611 Chr.8: 94260131-94260131 on Build GRCh38
rs775753253 Chr.8: 94260159-94260159 on Build GRCh38
rs527705763 Chr.8: 94260185-94260185 on Build GRCh38
rs764523532 Chr.8: 94260298-94260300 on Build GRCh38
rs145046153 Chr.8: 94260309-94260309 on Build GRCh38
rs548062745 Chr.8: 94260191-94260191 on Build GRCh38
rs758312817 Chr.8: 94260192-94260192 on Build GRCh38
rs567771198 Chr.8: 94260241-94260241 on Build GRCh38
rs201792479 Chr.8: 94260183-94260183 on Build GRCh38

View Full Product Details