Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34177741-34177992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf94
Entrez Gene ID
84970
Gene Name
chromosome 1 open reading frame 94
Gene Aliases
-
UniGene
Hs.194610
Gene Chromosome Location
Chr.1: 34166883-34219131 on Build GRCh38
Transcripts
NM_001134734.1 NM_032884.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5598071 c.132G>T p.L44L Chr.1: 34177921-34177921 on Build GRCh38
6042208 c.4A>G p.R2G Chr.1: 34177793-34177793 on Build GRCh38
6439502 c.7G>A p.G3S Chr.1: 34177796-34177796 on Build GRCh38
4752588 c.52C>T p.Q18* Chr.1: 34177841-34177841 on Build GRCh38
3977609 c.96A>G p.S32S Chr.1: 34177885-34177885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549437784 Chr.1: 34177794-34177794 on Build GRCh38
rs910107980 Chr.1: 34177959-34177959 on Build GRCh38
rs1021187066 Chr.1: 34177852-34177852 on Build GRCh38
rs755130288 Chr.1: 34177916-34177916 on Build GRCh38
rs1053329954 Chr.1: 34177867-34177867 on Build GRCh38
rs764776804 Chr.1: 34177849-34177851 on Build GRCh38
rs751844758 Chr.1: 34177914-34177914 on Build GRCh38
rs753245033 Chr.1: 34177763-34177763 on Build GRCh38
rs201119842 Chr.1: 34177964-34177964 on Build GRCh38
rs781422404 Chr.1: 34177920-34177920 on Build GRCh38
rs184414690 Chr.1: 34177812-34177812 on Build GRCh38
rs564391103 Chr.1: 34177963-34177963 on Build GRCh38
rs1043804215 Chr.1: 34177890-34177890 on Build GRCh38
rs575665507 Chr.1: 34177865-34177865 on Build GRCh38
rs892084382 Chr.1: 34177870-34177870 on Build GRCh38
rs749863622 Chr.1: 34177909-34177909 on Build GRCh38
rs775171633 Chr.1: 34177807-34177807 on Build GRCh38
rs767329804 Chr.1: 34177876-34177876 on Build GRCh38
rs980223350 Chr.1: 34177835-34177835 on Build GRCh38
rs983808627 Chr.1: 34177946-34177946 on Build GRCh38
rs760478579 Chr.1: 34177813-34177813 on Build GRCh38
rs764131580 Chr.1: 34177814-34177814 on Build GRCh38
rs546331689 Chr.1: 34177891-34177891 on Build GRCh38
rs753769207 Chr.1: 34177825-34177825 on Build GRCh38
rs41266411 Chr.1: 34177849-34177849 on Build GRCh38
rs940254429 Chr.1: 34177962-34177962 on Build GRCh38
rs756631480 Chr.1: 34177793-34177794 on Build GRCh38
rs766868298 Chr.1: 34177761-34177761 on Build GRCh38
rs921025180 Chr.1: 34177915-34177915 on Build GRCh38
rs775330338 Chr.1: 34177907-34177907 on Build GRCh38

View Full Product Details