Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5465414-5465683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD274
Entrez Gene ID
29126
Gene Name
CD274 molecule
Gene Aliases
B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1
UniGene
Hs.521989
Gene Chromosome Location
Chr.9: 5450503-5470567 on Build GRCh38
Transcripts
NM_001267706.1 NM_014143.3 NR_052005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
346133 c.726A>T p.V242V Chr.9: 5465542-5465542 on Build GRCh38
293117 c.778C>T p.R260C Chr.9: 5465594-5465594 on Build GRCh38
6959842 c.737C>G p.A246G Chr.9: 5465553-5465553 on Build GRCh38
5626330 c.683-1G>A p.? Chr.9: 5465498-5465498 on Build GRCh38
1247690 c.741C>A p.I247I Chr.9: 5465557-5465557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035717099 Chr.9: 5465586-5465586 on Build GRCh38
rs556678185 Chr.9: 5465440-5465440 on Build GRCh38
rs756183119 Chr.9: 5465485-5465485 on Build GRCh38
rs747218330 Chr.9: 5465511-5465511 on Build GRCh38
rs780617203 Chr.9: 5465637-5465637 on Build GRCh38
rs760272527 Chr.9: 5465590-5465590 on Build GRCh38
rs768935500 Chr.9: 5465546-5465546 on Build GRCh38
rs764989247 Chr.9: 5465615-5465615 on Build GRCh38
rs746369816 Chr.9: 5465525-5465525 on Build GRCh38
rs369938558 Chr.9: 5465480-5465480 on Build GRCh38
rs568608390 Chr.9: 5465594-5465594 on Build GRCh38
rs769528781 Chr.9: 5465479-5465480 on Build GRCh38
rs766260633 Chr.9: 5465490-5465490 on Build GRCh38
rs762887168 Chr.9: 5465602-5465602 on Build GRCh38
rs372727420 Chr.9: 5465530-5465530 on Build GRCh38
rs774011022 Chr.9: 5465550-5465550 on Build GRCh38
rs745892005 Chr.9: 5465653-5465653 on Build GRCh38
rs976620888 Chr.9: 5465478-5465478 on Build GRCh38
rs761351112 Chr.9: 5465479-5465479 on Build GRCh38
rs781235889 Chr.9: 5465651-5465651 on Build GRCh38
rs918780874 Chr.9: 5465652-5465652 on Build GRCh38
rs775247957 Chr.9: 5465558-5465560 on Build GRCh38
rs999341393 Chr.9: 5465452-5465452 on Build GRCh38
rs148141792 Chr.9: 5465595-5465595 on Build GRCh38
rs754751977 Chr.9: 5465596-5465596 on Build GRCh38
rs756192559 Chr.9: 5465658-5465658 on Build GRCh38
rs753626864 Chr.9: 5465496-5465496 on Build GRCh38
rs763747780 Chr.9: 5465468-5465468 on Build GRCh38
rs762582717 Chr.9: 5465463-5465463 on Build GRCh38
rs767045783 Chr.9: 5465569-5465569 on Build GRCh38
rs754823390 Chr.9: 5465497-5465497 on Build GRCh38
rs772911038 Chr.9: 5465572-5465572 on Build GRCh38
rs775991047 Chr.9: 5465535-5465535 on Build GRCh38
rs750352211 Chr.9: 5465483-5465483 on Build GRCh38
rs775900667 Chr.9: 5465449-5465449 on Build GRCh38
rs749591323 Chr.9: 5465542-5465542 on Build GRCh38
rs757488543 Chr.9: 5465650-5465650 on Build GRCh38
rs773882875 Chr.9: 5465470-5465470 on Build GRCh38
rs759252442 Chr.9: 5465471-5465471 on Build GRCh38
rs761425208 Chr.9: 5465552-5465552 on Build GRCh38
rs778013992 Chr.9: 5465498-5465498 on Build GRCh38
rs751773137 Chr.9: 5465632-5465632 on Build GRCh38
rs61752860 Chr.9: 5465514-5465514 on Build GRCh38
rs953036942 Chr.9: 5465660-5465660 on Build GRCh38
rs763972047 Chr.9: 5465611-5465612 on Build GRCh38
rs757337666 Chr.9: 5465513-5465513 on Build GRCh38
rs972846540 Chr.9: 5465606-5465607 on Build GRCh38

View Full Product Details