Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119586232-119586471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRH
Entrez Gene ID
141
Gene Name
ADP-ribosylarginine hydrolase
Gene Aliases
ARH1
UniGene
Hs.99884
Gene Chromosome Location
Chr.3: 119579433-119589954 on Build GRCh38
Transcripts
NM_001125.3 NM_001291949.1 NM_001291950.1 XM_011512438.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5584398 c.457C>T p.Q153* Chr.3: 119586443-119586443 on Build GRCh38
4760623 c.415G>A p.G139S Chr.3: 119586401-119586401 on Build GRCh38
341920 c.481A>G p.M161V Chr.3: 119586467-119586467 on Build GRCh38
6595345 c.381C>T p.G127G Chr.3: 119586367-119586367 on Build GRCh38
4112548 c.414C>T p.I138I Chr.3: 119586400-119586400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781238699 Chr.3: 119586304-119586304 on Build GRCh38
rs1042021663 Chr.3: 119586372-119586372 on Build GRCh38
rs368454824 Chr.3: 119586332-119586332 on Build GRCh38
rs746589265 Chr.3: 119586420-119586420 on Build GRCh38
rs763506189 Chr.3: 119586262-119586262 on Build GRCh38
rs142760415 Chr.3: 119586295-119586295 on Build GRCh38
rs536835076 Chr.3: 119586358-119586358 on Build GRCh38
rs751906391 Chr.3: 119586270-119586270 on Build GRCh38
rs20569 Chr.3: 119586322-119586322 on Build GRCh38
rs747882109 Chr.3: 119586301-119586301 on Build GRCh38
rs146226998 Chr.3: 119586364-119586364 on Build GRCh38
rs769932531 Chr.3: 119586313-119586313 on Build GRCh38
rs760314743 Chr.3: 119586351-119586351 on Build GRCh38
rs143159355 Chr.3: 119586284-119586284 on Build GRCh38
rs774972021 Chr.3: 119586395-119586395 on Build GRCh38
rs759014619 Chr.3: 119586425-119586425 on Build GRCh38
rs769283255 Chr.3: 119586328-119586328 on Build GRCh38
rs779491843 Chr.3: 119586333-119586333 on Build GRCh38
rs750142650 Chr.3: 119586277-119586277 on Build GRCh38
rs764684610 Chr.3: 119586439-119586439 on Build GRCh38
rs777573670 Chr.3: 119586368-119586368 on Build GRCh38
rs752720813 Chr.3: 119586365-119586365 on Build GRCh38
rs367735796 Chr.3: 119586294-119586294 on Build GRCh38
rs963707407 Chr.3: 119586302-119586302 on Build GRCh38
rs752168730 Chr.3: 119586447-119586447 on Build GRCh38
rs146369894 Chr.3: 119586400-119586400 on Build GRCh38
rs377275745 Chr.3: 119586355-119586355 on Build GRCh38
rs919242705 Chr.3: 119586424-119586424 on Build GRCh38
rs764713904 Chr.3: 119586265-119586265 on Build GRCh38
rs769954113 Chr.3: 119586257-119586257 on Build GRCh38
rs373614749 Chr.3: 119586271-119586271 on Build GRCh38
rs142179039 Chr.3: 119586387-119586387 on Build GRCh38
rs750963214 Chr.3: 119586256-119586256 on Build GRCh38
rs746061887 Chr.3: 119586308-119586308 on Build GRCh38
rs756340362 Chr.3: 119586375-119586375 on Build GRCh38
rs986549329 Chr.3: 119586288-119586288 on Build GRCh38
rs137924029 Chr.3: 119586386-119586386 on Build GRCh38
rs779502800 Chr.3: 119586280-119586280 on Build GRCh38
rs766021832 Chr.3: 119586354-119586354 on Build GRCh38
rs748650536 Chr.3: 119586398-119586398 on Build GRCh38
rs775655162 Chr.3: 119586318-119586318 on Build GRCh38
rs761010216 Chr.3: 119586401-119586401 on Build GRCh38
rs1020589398 Chr.3: 119586292-119586292 on Build GRCh38
rs555140584 Chr.3: 119586367-119586367 on Build GRCh38
rs778885533 Chr.3: 119586300-119586300 on Build GRCh38
rs138814998 Chr.3: 119586279-119586279 on Build GRCh38
rs759512476 Chr.3: 119586254-119586254 on Build GRCh38
rs750787316 Chr.3: 119586341-119586341 on Build GRCh38
rs780491592 Chr.3: 119586383-119586383 on Build GRCh38
rs369779554 Chr.3: 119586369-119586369 on Build GRCh38
rs767066865 Chr.3: 119586272-119586272 on Build GRCh38

View Full Product Details