Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 60742197-60742422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD7
Entrez Gene ID
55636
Gene Name
chromodomain helicase DNA binding protein 7
Gene Aliases
CRG, HH5, IS3, KAL5
UniGene
Hs.20395
Gene Chromosome Location
Chr.8: 60678744-60868028 on Build GRCh38
Transcripts
NM_001316690.1 NM_017780.3 XM_011517553.2 XM_011517554.2 XM_011517555.2 XM_011517560.2 XM_017013612.1 XM_017013613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3900854 c.779C>T p.P260L Chr.8: 60742211-60742211 on Build GRCh38
6628146 c.925C>T p.Q309* Chr.8: 60742357-60742357 on Build GRCh38
4989042 c.805G>A p.V269I Chr.8: 60742237-60742237 on Build GRCh38
2719979 c.884G>A p.R295Q Chr.8: 60742316-60742316 on Build GRCh38
5584788 c.899C>T p.P300L Chr.8: 60742331-60742331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775415630 Chr.8: 60742263-60742263 on Build GRCh38
rs777428817 Chr.8: 60742359-60742359 on Build GRCh38
rs749302988 Chr.8: 60742379-60742379 on Build GRCh38
rs376743531 Chr.8: 60742336-60742336 on Build GRCh38
rs768312325 Chr.8: 60742233-60742233 on Build GRCh38
rs747972265 Chr.8: 60742237-60742237 on Build GRCh38
rs747781367 Chr.8: 60742339-60742339 on Build GRCh38
rs13248429 Chr.8: 60742360-60742360 on Build GRCh38
rs774274534 Chr.8: 60742384-60742384 on Build GRCh38
rs878891577 Chr.8: 60742350-60742350 on Build GRCh38
rs13248404 Chr.8: 60742328-60742328 on Build GRCh38
rs746735047 Chr.8: 60742232-60742232 on Build GRCh38
rs886041166 Chr.8: 60742366-60742366 on Build GRCh38
rs61995713 Chr.8: 60742288-60742288 on Build GRCh38
rs780756877 Chr.8: 60742236-60742236 on Build GRCh38
rs530158598 Chr.8: 60742243-60742243 on Build GRCh38
rs760775347 Chr.8: 60742271-60742271 on Build GRCh38
rs992508075 Chr.8: 60742260-60742260 on Build GRCh38
rs955811265 Chr.8: 60742367-60742367 on Build GRCh38
rs1010032031 Chr.8: 60742378-60742378 on Build GRCh38
rs779427012 Chr.8: 60742226-60742226 on Build GRCh38
rs762771594 Chr.8: 60742250-60742250 on Build GRCh38
rs764411605 Chr.8: 60742275-60742275 on Build GRCh38
rs772257683 Chr.8: 60742262-60742262 on Build GRCh38
rs770785044 Chr.8: 60742383-60742383 on Build GRCh38
rs537091457 Chr.8: 60742343-60742343 on Build GRCh38
rs765674058 Chr.8: 60742303-60742303 on Build GRCh38
rs1007919674 Chr.8: 60742293-60742293 on Build GRCh38
rs766853484 Chr.8: 60742317-60742317 on Build GRCh38
rs750978197 Chr.8: 60742315-60742315 on Build GRCh38
rs754445442 Chr.8: 60742320-60742320 on Build GRCh38
rs569799707 Chr.8: 60742316-60742316 on Build GRCh38
rs773253450 Chr.8: 60742245-60742245 on Build GRCh38
rs886063033 Chr.8: 60742324-60742324 on Build GRCh38

View Full Product Details