Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38384343-38384596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGN
Entrez Gene ID
199720
Gene Name
gametogenetin
Gene Aliases
-
UniGene
Hs.447948
Gene Chromosome Location
Chr.19: 38384347-38388603 on Build GRCh38
Transcripts
NM_152657.3 XM_005258619.4 XM_011526603.2 XM_017026451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1680986 c.1939C>T p.Q647* Chr.19: 38384432-38384432 on Build GRCh38
1680985 c.1939C>G p.Q647E Chr.19: 38384432-38384432 on Build GRCh38
1208145 c.1876C>T p.P626S Chr.19: 38384495-38384495 on Build GRCh38
5750318 c.1933G>T p.D645Y Chr.19: 38384438-38384438 on Build GRCh38
7412904 c.1862A>G p.H621R Chr.19: 38384509-38384509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769894210 Chr.19: 38384383-38384383 on Build GRCh38
rs778727013 Chr.19: 38384541-38384541 on Build GRCh38
rs754923601 Chr.19: 38384536-38384536 on Build GRCh38
rs200653001 Chr.19: 38384469-38384469 on Build GRCh38
rs747801740 Chr.19: 38384471-38384471 on Build GRCh38
rs780069031 Chr.19: 38384533-38384533 on Build GRCh38
rs369255044 Chr.19: 38384389-38384389 on Build GRCh38
rs781391103 Chr.19: 38384439-38384439 on Build GRCh38
rs759993569 Chr.19: 38384424-38384424 on Build GRCh38
rs776060798 Chr.19: 38384505-38384505 on Build GRCh38
rs760187766 Chr.19: 38384501-38384501 on Build GRCh38
rs369901778 Chr.19: 38384468-38384468 on Build GRCh38
rs746206553 Chr.19: 38384378-38384378 on Build GRCh38
rs148924874 Chr.19: 38384467-38384467 on Build GRCh38
rs917110030 Chr.19: 38384456-38384456 on Build GRCh38
rs750473087 Chr.19: 38384528-38384528 on Build GRCh38
rs755182284 Chr.19: 38384545-38384546 on Build GRCh38
rs764495999 Chr.19: 38384517-38384517 on Build GRCh38
rs747957661 Chr.19: 38384544-38384544 on Build GRCh38
rs200897385 Chr.19: 38384403-38384403 on Build GRCh38
rs763425580 Chr.19: 38384512-38384512 on Build GRCh38
rs200182647 Chr.19: 38384401-38384401 on Build GRCh38
rs781478900 Chr.19: 38384373-38384373 on Build GRCh38
rs890622869 Chr.19: 38384372-38384372 on Build GRCh38
rs964629029 Chr.19: 38384566-38384566 on Build GRCh38
rs774075127 Chr.19: 38384470-38384470 on Build GRCh38
rs770414689 Chr.19: 38384568-38384568 on Build GRCh38
rs372987753 Chr.19: 38384404-38384404 on Build GRCh38
rs775385331 Chr.19: 38384386-38384386 on Build GRCh38
rs757352062 Chr.19: 38384369-38384369 on Build GRCh38
rs765751420 Chr.19: 38384429-38384429 on Build GRCh38
rs541323480 Chr.19: 38384502-38384502 on Build GRCh38
rs768140126 Chr.19: 38384397-38384397 on Build GRCh38
rs201908140 Chr.19: 38384377-38384377 on Build GRCh38
rs139780646 Chr.19: 38384418-38384418 on Build GRCh38
rs374894863 Chr.19: 38384376-38384376 on Build GRCh38
rs377542328 Chr.19: 38384532-38384532 on Build GRCh38
rs777403768 Chr.19: 38384560-38384560 on Build GRCh38
rs745977429 Chr.19: 38384443-38384443 on Build GRCh38
rs776052948 Chr.19: 38384576-38384576 on Build GRCh38
rs767875539 Chr.19: 38384525-38384525 on Build GRCh38
rs752943142 Chr.19: 38384431-38384431 on Build GRCh38
rs145910727 Chr.19: 38384519-38384519 on Build GRCh38
rs772656083 Chr.19: 38384488-38384488 on Build GRCh38
rs977629443 Chr.19: 38384482-38384482 on Build GRCh38
rs771741257 Chr.19: 38384486-38384486 on Build GRCh38
rs1010288305 Chr.19: 38384450-38384450 on Build GRCh38
rs145717888 Chr.19: 38384438-38384438 on Build GRCh38
rs62123481 Chr.19: 38384432-38384432 on Build GRCh38
rs754093463 Chr.19: 38384481-38384493 on Build GRCh38
rs753803222 Chr.19: 38384535-38384535 on Build GRCh38
rs771651294 Chr.19: 38384557-38384557 on Build GRCh38

View Full Product Details