Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94567181-94567454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBP2
Entrez Gene ID
8789
Gene Name
fructose-bisphosphatase 2
Gene Aliases
-
UniGene
Hs.61255
Gene Chromosome Location
Chr.9: 94558714-94593832 on Build GRCh38
Transcripts
NM_003837.3 NR_121566.1 NR_121567.1 NR_121568.1 NR_121569.1

Gene Symbol
PCAT7
Entrez Gene ID
101928099
Gene Name
prostate cancer associated transcript 7 (non-protein coding)
Gene Aliases
PCAN-R2
UniGene
Hs.674409
Gene Chromosome Location
Chr.9: 94555045-94568141 on Build GRCh38
Transcripts
NM_003837.3 NR_121566.1 NR_121567.1 NR_121568.1 NR_121569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1206755 c.693G>T p.K231N Chr.9: 94567282-94567282 on Build GRCh38
5601475 c.644G>A p.G215D Chr.9: 94567331-94567331 on Build GRCh38
2732885 c.640G>C p.E214Q Chr.9: 94567335-94567335 on Build GRCh38
6442309 c.592G>A p.E198K Chr.9: 94567383-94567383 on Build GRCh38
1252228 c.682G>A p.V228M Chr.9: 94567293-94567293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767447027 Chr.9: 94567337-94567337 on Build GRCh38
rs763218467 Chr.9: 94567326-94567326 on Build GRCh38
rs761915951 Chr.9: 94567329-94567329 on Build GRCh38
rs757198213 Chr.9: 94567359-94567359 on Build GRCh38
rs746591066 Chr.9: 94567403-94567403 on Build GRCh38
rs202197694 Chr.9: 94567423-94567423 on Build GRCh38
rs946948601 Chr.9: 94567225-94567225 on Build GRCh38
rs546385370 Chr.9: 94567315-94567315 on Build GRCh38
rs548635230 Chr.9: 94567345-94567345 on Build GRCh38
rs764653047 Chr.9: 94567226-94567226 on Build GRCh38
rs201631172 Chr.9: 94567272-94567272 on Build GRCh38
rs780120443 Chr.9: 94567251-94567251 on Build GRCh38
rs201102588 Chr.9: 94567420-94567420 on Build GRCh38
rs112480079 Chr.9: 94567305-94567305 on Build GRCh38
rs765532722 Chr.9: 94567236-94567236 on Build GRCh38
rs751644073 Chr.9: 94567255-94567255 on Build GRCh38
rs1040462521 Chr.9: 94567285-94567285 on Build GRCh38
rs373644937 Chr.9: 94567417-94567417 on Build GRCh38
rs770271913 Chr.9: 94567425-94567425 on Build GRCh38
rs978353233 Chr.9: 94567428-94567428 on Build GRCh38
rs756071228 Chr.9: 94567256-94567256 on Build GRCh38
rs778868837 Chr.9: 94567368-94567368 on Build GRCh38
rs779731457 Chr.9: 94567392-94567392 on Build GRCh38
rs770872181 Chr.9: 94567287-94567287 on Build GRCh38
rs376612944 Chr.9: 94567295-94567295 on Build GRCh38
rs755065804 Chr.9: 94567334-94567334 on Build GRCh38
rs374994599 Chr.9: 94567243-94567243 on Build GRCh38
rs111617337 Chr.9: 94567300-94567300 on Build GRCh38
rs1034752573 Chr.9: 94567365-94567365 on Build GRCh38
rs1003272425 Chr.9: 94567364-94567364 on Build GRCh38
rs7036264 Chr.9: 94567259-94567259 on Build GRCh38
rs762140370 Chr.9: 94567228-94567228 on Build GRCh38
rs778811613 Chr.9: 94567297-94567297 on Build GRCh38
rs141540383 Chr.9: 94567377-94567377 on Build GRCh38
rs915508652 Chr.9: 94567203-94567203 on Build GRCh38
rs758195538 Chr.9: 94567383-94567383 on Build GRCh38
rs61755092 Chr.9: 94567343-94567343 on Build GRCh38
rs113040440 Chr.9: 94567266-94567266 on Build GRCh38
rs367608023 Chr.9: 94567309-94567309 on Build GRCh38
rs944711585 Chr.9: 94567390-94567390 on Build GRCh38
rs754191275 Chr.9: 94567227-94567227 on Build GRCh38
rs768240058 Chr.9: 94567406-94567406 on Build GRCh38
rs959088264 Chr.9: 94567372-94567372 on Build GRCh38
rs774922595 Chr.9: 94567304-94567304 on Build GRCh38
rs765587674 Chr.9: 94567321-94567321 on Build GRCh38
rs1000204080 Chr.9: 94567204-94567204 on Build GRCh38
rs371546490 Chr.9: 94567310-94567310 on Build GRCh38
rs372051710 Chr.9: 94567239-94567239 on Build GRCh38
rs146159894 Chr.9: 94567328-94567328 on Build GRCh38

View Full Product Details