Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94536020-94536267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F3
Entrez Gene ID
2152
Gene Name
coagulation factor III, tissue factor
Gene Aliases
CD142, TF, TFA
UniGene
Hs.62192
Gene Chromosome Location
Chr.1: 94529176-94541857 on Build GRCh38
Transcripts
NM_001178096.1 NM_001993.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4400441 c.281A>G p.E94G Chr.1: 94536096-94536096 on Build GRCh38
5530725 c.280G>A p.E94K Chr.1: 94536097-94536097 on Build GRCh38
6657736 c.332C>T p.P111L Chr.1: 94536045-94536045 on Build GRCh38
426871 c.354C>A p.T118T Chr.1: 94536023-94536023 on Build GRCh38
913513 c.242G>T p.C81F Chr.1: 94536135-94536135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747397927 Chr.1: 94536061-94536061 on Build GRCh38
rs908328573 Chr.1: 94536182-94536182 on Build GRCh38
rs558602649 Chr.1: 94536196-94536196 on Build GRCh38
rs371434099 Chr.1: 94536072-94536072 on Build GRCh38
rs777936997 Chr.1: 94536045-94536045 on Build GRCh38
rs767254897 Chr.1: 94536186-94536186 on Build GRCh38
rs116345246 Chr.1: 94536222-94536222 on Build GRCh38
rs989914810 Chr.1: 94536071-94536071 on Build GRCh38
rs569628976 Chr.1: 94536100-94536100 on Build GRCh38
rs765245806 Chr.1: 94536206-94536206 on Build GRCh38
rs77418724 Chr.1: 94536126-94536126 on Build GRCh38
rs748868875 Chr.1: 94536181-94536181 on Build GRCh38
rs764859311 Chr.1: 94536154-94536157 on Build GRCh38
rs377083934 Chr.1: 94536056-94536056 on Build GRCh38
rs199905967 Chr.1: 94536097-94536097 on Build GRCh38
rs370240526 Chr.1: 94536202-94536202 on Build GRCh38
rs922921059 Chr.1: 94536113-94536113 on Build GRCh38
rs534782791 Chr.1: 94536225-94536225 on Build GRCh38
rs943081176 Chr.1: 94536080-94536080 on Build GRCh38
rs745543730 Chr.1: 94536089-94536089 on Build GRCh38
rs929612920 Chr.1: 94536123-94536123 on Build GRCh38
rs780973921 Chr.1: 94536076-94536076 on Build GRCh38
rs924494158 Chr.1: 94536051-94536051 on Build GRCh38
rs769375517 Chr.1: 94536092-94536092 on Build GRCh38
rs13306323 Chr.1: 94536217-94536217 on Build GRCh38
rs777105331 Chr.1: 94536101-94536101 on Build GRCh38
rs143237190 Chr.1: 94536099-94536099 on Build GRCh38
rs957191238 Chr.1: 94536063-94536063 on Build GRCh38
rs698946 Chr.1: 94536044-94536044 on Build GRCh38
rs374919018 Chr.1: 94536163-94536163 on Build GRCh38
rs756646702 Chr.1: 94536076-94536077 on Build GRCh38
rs554918363 Chr.1: 94536231-94536231 on Build GRCh38
rs775151591 Chr.1: 94536098-94536098 on Build GRCh38
rs774211895 Chr.1: 94536152-94536152 on Build GRCh38

View Full Product Details