Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183319370-183319639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN22
Entrez Gene ID
53842
Gene Name
claudin 22
Gene Aliases
CLDN21
UniGene
Hs.333179
Gene Chromosome Location
Chr.4: 183318067-183320774 on Build GRCh38
Transcripts
NM_001111319.1 NM_024949.5

Gene Symbol
WWC2
Entrez Gene ID
80014
Gene Name
WW and C2 domain containing 2
Gene Aliases
BOMB
UniGene
Hs.333179
Gene Chromosome Location
Chr.4: 183098677-183320777 on Build GRCh38
Transcripts
NM_001111319.1 NM_024949.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7110063 c.638C>A p.T213K Chr.4: 183319581-183319581 on Build GRCh38
1428804 c.585T>G p.A195A Chr.4: 183319634-183319634 on Build GRCh38
4123644 c.582A>G p.L194L Chr.4: 183319637-183319637 on Build GRCh38
1485861 c.613C>A p.Q205K Chr.4: 183319606-183319606 on Build GRCh38
6630012 c.645C>T p.N215N Chr.4: 183319574-183319574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762242510 Chr.4: 183319603-183319603 on Build GRCh38
rs895345034 Chr.4: 183319497-183319497 on Build GRCh38
rs373938350 Chr.4: 183319588-183319588 on Build GRCh38
rs367950585 Chr.4: 183319558-183319558 on Build GRCh38
rs540564854 Chr.4: 183319539-183319539 on Build GRCh38
rs897917624 Chr.4: 183319436-183319436 on Build GRCh38
rs374445616 Chr.4: 183319510-183319510 on Build GRCh38
rs1048333093 Chr.4: 183319406-183319406 on Build GRCh38
rs1006638260 Chr.4: 183319422-183319422 on Build GRCh38
rs184295038 Chr.4: 183319420-183319420 on Build GRCh38
rs1030486601 Chr.4: 183319445-183319445 on Build GRCh38
rs933289149 Chr.4: 183319583-183319583 on Build GRCh38
rs769359354 Chr.4: 183319605-183319605 on Build GRCh38
rs201257600 Chr.4: 183319522-183319522 on Build GRCh38
rs772302316 Chr.4: 183319464-183319464 on Build GRCh38
rs371522285 Chr.4: 183319572-183319572 on Build GRCh38
rs923264814 Chr.4: 183319421-183319421 on Build GRCh38
rs771779883 Chr.4: 183319489-183319489 on Build GRCh38
rs897849368 Chr.4: 183319604-183319604 on Build GRCh38
rs372301775 Chr.4: 183319524-183319524 on Build GRCh38
rs201506772 Chr.4: 183319581-183319581 on Build GRCh38
rs750082273 Chr.4: 183319521-183319521 on Build GRCh38
rs761336229 Chr.4: 183319513-183319513 on Build GRCh38
rs751476729 Chr.4: 183319426-183319426 on Build GRCh38
rs777137443 Chr.4: 183319594-183319594 on Build GRCh38
rs1022753063 Chr.4: 183319509-183319509 on Build GRCh38
rs866834887 Chr.4: 183319615-183319615 on Build GRCh38
rs752629967 Chr.4: 183319544-183319544 on Build GRCh38
rs751347206 Chr.4: 183319528-183319528 on Build GRCh38
rs755878121 Chr.4: 183319593-183319595 on Build GRCh38
rs745529424 Chr.4: 183319482-183319482 on Build GRCh38
rs1016045358 Chr.4: 183319547-183319547 on Build GRCh38
rs544421553 Chr.4: 183319606-183319606 on Build GRCh38
rs764670745 Chr.4: 183319515-183319515 on Build GRCh38
rs573875393 Chr.4: 183319450-183319450 on Build GRCh38
rs762561673 Chr.4: 183319613-183319613 on Build GRCh38
rs978137056 Chr.4: 183319587-183319587 on Build GRCh38
rs562903430 Chr.4: 183319621-183319621 on Build GRCh38
rs1039130761 Chr.4: 183319416-183319416 on Build GRCh38
rs371516258 Chr.4: 183319507-183319507 on Build GRCh38
rs997628447 Chr.4: 183319437-183319437 on Build GRCh38
rs770878452 Chr.4: 183319412-183319412 on Build GRCh38
rs199788573 Chr.4: 183319540-183319540 on Build GRCh38
rs757422855 Chr.4: 183319550-183319550 on Build GRCh38
rs766023424 Chr.4: 183319617-183319617 on Build GRCh38
rs1014986815 Chr.4: 183319424-183319428 on Build GRCh38
rs964556368 Chr.4: 183319563-183319563 on Build GRCh38
rs986371714 Chr.4: 183319476-183319476 on Build GRCh38
rs772319025 Chr.4: 183319580-183319580 on Build GRCh38
rs535186367 Chr.4: 183319488-183319488 on Build GRCh38
rs1012662904 Chr.4: 183319499-183319499 on Build GRCh38
rs933338590 Chr.4: 183319438-183319438 on Build GRCh38

View Full Product Details