Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12719323-12719588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADACL3
Entrez Gene ID
126767
Gene Name
arylacetamide deacetylase like 3
Gene Aliases
-
UniGene
Hs.464705
Gene Chromosome Location
Chr.1: 12716115-12728759 on Build GRCh38
Transcripts
NM_001103170.2 NR_111984.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1333380 c.106C>T p.R36C Chr.1: 12719583-12719583 on Build GRCh38
1333379 c.98C>T p.T33M Chr.1: 12719575-12719575 on Build GRCh38
5377459 c.3G>A p.M1I Chr.1: 12719480-12719480 on Build GRCh38
894877 c.12G>T p.E4D Chr.1: 12719489-12719489 on Build GRCh38
6057746 c.55C>T p.Q19* Chr.1: 12719532-12719532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78526061 Chr.1: 12719562-12719562 on Build GRCh38
rs372017787 Chr.1: 12719462-12719462 on Build GRCh38
rs896528411 Chr.1: 12719497-12719497 on Build GRCh38
rs3010876 Chr.1: 12719558-12719558 on Build GRCh38
rs748421323 Chr.1: 12719554-12719554 on Build GRCh38
rs941571131 Chr.1: 12719402-12719402 on Build GRCh38
rs549280967 Chr.1: 12719368-12719368 on Build GRCh38
rs964086085 Chr.1: 12719416-12719416 on Build GRCh38
rs756482950 Chr.1: 12719540-12719540 on Build GRCh38
rs751162776 Chr.1: 12719538-12719538 on Build GRCh38
rs181866526 Chr.1: 12719464-12719464 on Build GRCh38
rs761961725 Chr.1: 12719523-12719523 on Build GRCh38
rs762633784 Chr.1: 12719516-12719516 on Build GRCh38
rs747151765 Chr.1: 12719417-12719417 on Build GRCh38
rs377178173 Chr.1: 12719547-12719547 on Build GRCh38
rs1047135769 Chr.1: 12719391-12719391 on Build GRCh38
rs747553258 Chr.1: 12719418-12719418 on Build GRCh38
rs142242506 Chr.1: 12719545-12719545 on Build GRCh38
rs367966099 Chr.1: 12719447-12719447 on Build GRCh38
rs201816325 Chr.1: 12719561-12719562 on Build GRCh38
rs926639988 Chr.1: 12719536-12719536 on Build GRCh38
rs751789343 Chr.1: 12719521-12719522 on Build GRCh38
rs371665362 Chr.1: 12719546-12719546 on Build GRCh38
rs776633019 Chr.1: 12719466-12719466 on Build GRCh38
rs752123416 Chr.1: 12719510-12719510 on Build GRCh38
rs774289955 Chr.1: 12719443-12719443 on Build GRCh38
rs974430242 Chr.1: 12719502-12719502 on Build GRCh38
rs749701061 Chr.1: 12719432-12719432 on Build GRCh38
rs933665788 Chr.1: 12719385-12719385 on Build GRCh38
rs200596255 Chr.1: 12719543-12719543 on Build GRCh38
rs115702857 Chr.1: 12719400-12719400 on Build GRCh38
rs773185052 Chr.1: 12719451-12719451 on Build GRCh38
rs184851056 Chr.1: 12719475-12719475 on Build GRCh38
rs767588489 Chr.1: 12719566-12719566 on Build GRCh38
rs999824593 Chr.1: 12719361-12719361 on Build GRCh38
rs780218262 Chr.1: 12719541-12719541 on Build GRCh38
rs560269535 Chr.1: 12719351-12719351 on Build GRCh38
rs945504840 Chr.1: 12719551-12719551 on Build GRCh38
rs768787814 Chr.1: 12719436-12719436 on Build GRCh38
rs1056544689 Chr.1: 12719474-12719474 on Build GRCh38
rs908425822 Chr.1: 12719450-12719450 on Build GRCh38
rs529205133 Chr.1: 12719353-12719353 on Build GRCh38
rs982595404 Chr.1: 12719449-12719449 on Build GRCh38
rs78912772 Chr.1: 12719561-12719561 on Build GRCh38
rs755218857 Chr.1: 12719446-12719446 on Build GRCh38
rs376918948 Chr.1: 12719529-12719529 on Build GRCh38
rs370140994 Chr.1: 12719552-12719552 on Build GRCh38
rs772243495 Chr.1: 12719555-12719555 on Build GRCh38

View Full Product Details