Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176336523-176336796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIMC1
Entrez Gene ID
375484
Gene Name
SUMO interacting motifs containing 1
Gene Aliases
C5orf25, OOMA1, PLEIAD
UniGene
Hs.719847
Gene Chromosome Location
Chr.5: 176238359-176345991 on Build GRCh38
Transcripts
NM_001308195.1 NM_001308196.1 NM_001308200.1 NM_198567.5 NR_131772.1 XM_011534553.2 XM_011534554.2 XM_011534556.2 XM_017009454.1 XM_017009455.1 XM_017009456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1065990 c.2124C>A p.F708L Chr.5: 176336729-176336729 on Build GRCh38
1065989 c.2116G>T p.E706* Chr.5: 176336721-176336721 on Build GRCh38
3614299 c.501A>T p.E167D Chr.5: 176336723-176336723 on Build GRCh38
1733617 c.2167G>A p.E723K Chr.5: 176336772-176336772 on Build GRCh38
3614300 c.2155T>A p.C719S Chr.5: 176336760-176336760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541288445 Chr.5: 176336740-176336740 on Build GRCh38
rs760903170 Chr.5: 176336758-176336758 on Build GRCh38
rs762128351 Chr.5: 176336565-176336565 on Build GRCh38
rs1040197854 Chr.5: 176336624-176336624 on Build GRCh38
rs1029698727 Chr.5: 176336663-176336663 on Build GRCh38
rs773008022 Chr.5: 176336716-176336716 on Build GRCh38
rs375524685 Chr.5: 176336599-176336599 on Build GRCh38
rs116046473 Chr.5: 176336594-176336594 on Build GRCh38
rs753899816 Chr.5: 176336766-176336766 on Build GRCh38
rs746515664 Chr.5: 176336674-176336674 on Build GRCh38
rs35111138 Chr.5: 176336598-176336599 on Build GRCh38
rs527350811 Chr.5: 176336615-176336615 on Build GRCh38
rs754121478 Chr.5: 176336769-176336769 on Build GRCh38
rs965101004 Chr.5: 176336734-176336734 on Build GRCh38
rs748051323 Chr.5: 176336685-176336685 on Build GRCh38
rs897335679 Chr.5: 176336634-176336634 on Build GRCh38
rs772248367 Chr.5: 176336725-176336725 on Build GRCh38
rs780579908 Chr.5: 176336683-176336683 on Build GRCh38
rs762781110 Chr.5: 176336718-176336718 on Build GRCh38
rs867874942 Chr.5: 176336639-176336639 on Build GRCh38
rs564067290 Chr.5: 176336757-176336757 on Build GRCh38
rs377642720 Chr.5: 176336678-176336678 on Build GRCh38
rs148685734 Chr.5: 176336660-176336660 on Build GRCh38
rs769649449 Chr.5: 176336697-176336697 on Build GRCh38
rs892455497 Chr.5: 176336703-176336703 on Build GRCh38
rs779894520 Chr.5: 176336673-176336673 on Build GRCh38
rs143629670 Chr.5: 176336762-176336762 on Build GRCh38
rs67874748 Chr.5: 176336593-176336593 on Build GRCh38

View Full Product Details