Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35778977-35779230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP33
Entrez Gene ID
115703
Gene Name
Rho GTPase activating protein 33
Gene Aliases
NOMA-GAP, SNX26, TCGAP
UniGene
Hs.515364
Gene Chromosome Location
Chr.19: 35775515-35788822 on Build GRCh38
Transcripts
NM_001172630.1 NM_052948.3 XM_005258488.1 XM_006722999.1 XM_011526416.1 XM_011526417.1 XM_011526418.1 XM_011526419.1 XM_011526420.1 XM_011526421.2 XM_017026238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5674842 c.478G>T p.G160W Chr.19: 35779101-35779101 on Build GRCh38
5674844 c.532G>T p.G178W Chr.19: 35779101-35779101 on Build GRCh38
5163571 c.44G>A p.G15E Chr.19: 35779075-35779075 on Build GRCh38
5674843 c.70G>T p.G24W Chr.19: 35779101-35779101 on Build GRCh38
5163572 c.506G>A p.G169E Chr.19: 35779075-35779075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs231231 Chr.19: 35779150-35779150 on Build GRCh38
rs959723516 Chr.19: 35779075-35779075 on Build GRCh38
rs758583349 Chr.19: 35779025-35779025 on Build GRCh38
rs771324898 Chr.19: 35779195-35779196 on Build GRCh38
rs769821012 Chr.19: 35779139-35779139 on Build GRCh38
rs774164094 Chr.19: 35779165-35779165 on Build GRCh38
rs905583444 Chr.19: 35779017-35779017 on Build GRCh38
rs1021665784 Chr.19: 35779119-35779119 on Build GRCh38
rs377505912 Chr.19: 35779100-35779100 on Build GRCh38
rs778947202 Chr.19: 35779011-35779011 on Build GRCh38
rs781116109 Chr.19: 35779102-35779102 on Build GRCh38
rs761038358 Chr.19: 35779181-35779181 on Build GRCh38
rs1053659567 Chr.19: 35779014-35779014 on Build GRCh38
rs370580803 Chr.19: 35779142-35779142 on Build GRCh38
rs746769659 Chr.19: 35779039-35779039 on Build GRCh38
rs745787961 Chr.19: 35779138-35779138 on Build GRCh38
rs778635621 Chr.19: 35779008-35779008 on Build GRCh38
rs755578106 Chr.19: 35779010-35779010 on Build GRCh38
rs777241050 Chr.19: 35779196-35779197 on Build GRCh38
rs893620686 Chr.19: 35779026-35779026 on Build GRCh38
rs1012072541 Chr.19: 35779044-35779044 on Build GRCh38
rs231230 Chr.19: 35779013-35779013 on Build GRCh38
rs568132967 Chr.19: 35778998-35778998 on Build GRCh38
rs113013816 Chr.19: 35779155-35779155 on Build GRCh38
rs75900693 Chr.19: 35779206-35779206 on Build GRCh38
rs373417733 Chr.19: 35779086-35779086 on Build GRCh38
rs1001306726 Chr.19: 35779035-35779035 on Build GRCh38
rs768374643 Chr.19: 35779160-35779160 on Build GRCh38
rs770644481 Chr.19: 35779043-35779043 on Build GRCh38
rs777998253 Chr.19: 35779038-35779038 on Build GRCh38

View Full Product Details