Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39505058-39505319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLL3
Entrez Gene ID
10683
Gene Name
delta like canonical Notch ligand 3
Gene Aliases
SCDO1
UniGene
Hs.127792
Gene Chromosome Location
Chr.19: 39498917-39508481 on Build GRCh38
Transcripts
NM_016941.3 NM_203486.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5605675 c.901C>T p.P301S Chr.19: 39505259-39505259 on Build GRCh38
7122781 c.952G>C p.D318H Chr.19: 39505310-39505310 on Build GRCh38
4282716 c.928G>C p.E310Q Chr.19: 39505286-39505286 on Build GRCh38
3823053 c.904C>T p.R302C Chr.19: 39505262-39505262 on Build GRCh38
4078013 c.890A>T p.E297V Chr.19: 39505248-39505248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201781799 Chr.19: 39505219-39505219 on Build GRCh38
rs747538194 Chr.19: 39505236-39505236 on Build GRCh38
rs1046703236 Chr.19: 39505266-39505266 on Build GRCh38
rs753915676 Chr.19: 39505254-39505254 on Build GRCh38
rs773079824 Chr.19: 39505288-39505288 on Build GRCh38
rs755016225 Chr.19: 39505219-39505219 on Build GRCh38
rs750118522 Chr.19: 39505198-39505198 on Build GRCh38
rs753375994 Chr.19: 39505209-39505209 on Build GRCh38
rs778193263 Chr.19: 39505223-39505223 on Build GRCh38
rs537538122 Chr.19: 39505203-39505203 on Build GRCh38
rs986849875 Chr.19: 39505174-39505174 on Build GRCh38
rs577477375 Chr.19: 39505181-39505181 on Build GRCh38
rs757634002 Chr.19: 39505248-39505248 on Build GRCh38
rs553945114 Chr.19: 39505277-39505277 on Build GRCh38
rs7249157 Chr.19: 39505270-39505270 on Build GRCh38
rs753670322 Chr.19: 39505180-39505181 on Build GRCh38
rs755777033 Chr.19: 39505201-39505201 on Build GRCh38
rs767335941 Chr.19: 39505197-39505197 on Build GRCh38
rs141671275 Chr.19: 39505281-39505281 on Build GRCh38
rs148016836 Chr.19: 39505261-39505261 on Build GRCh38
rs534524753 Chr.19: 39505088-39505088 on Build GRCh38
rs560089545 Chr.19: 39505155-39505155 on Build GRCh38
rs372702131 Chr.19: 39505273-39505273 on Build GRCh38
rs572215947 Chr.19: 39505295-39505295 on Build GRCh38
rs988815394 Chr.19: 39505106-39505106 on Build GRCh38
rs766060828 Chr.19: 39505294-39505294 on Build GRCh38
rs377256847 Chr.19: 39505129-39505129 on Build GRCh38
rs150100958 Chr.19: 39505297-39505297 on Build GRCh38
rs778821687 Chr.19: 39505273-39505279 on Build GRCh38
rs760420806 Chr.19: 39505291-39505291 on Build GRCh38
rs919839538 Chr.19: 39505238-39505238 on Build GRCh38
rs3816049 Chr.19: 39505089-39505089 on Build GRCh38
rs955462291 Chr.19: 39505170-39505170 on Build GRCh38
rs781626135 Chr.19: 39505260-39505260 on Build GRCh38
rs761568223 Chr.19: 39505188-39505188 on Build GRCh38
rs749425668 Chr.19: 39505267-39505267 on Build GRCh38
rs775820811 Chr.19: 39505263-39505263 on Build GRCh38
rs774218328 Chr.19: 39505274-39505274 on Build GRCh38
rs538488581 Chr.19: 39505262-39505262 on Build GRCh38

View Full Product Details