Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85754778-85755015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATOH8
Entrez Gene ID
84913
Gene Name
atonal bHLH transcription factor 8
Gene Aliases
HATH6, bHLHa21
UniGene
Hs.135569
Gene Chromosome Location
Chr.2: 85753786-85791383 on Build GRCh38
Transcripts
NM_032827.6 XM_006712122.3 XM_011533139.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1023404 c.605A>G p.N202S Chr.2: 85754794-85754794 on Build GRCh38
3583780 c.734C>A p.T245N Chr.2: 85754923-85754923 on Build GRCh38
460986 c.751G>C p.E251Q Chr.2: 85754940-85754940 on Build GRCh38
7099382 c.708C>T p.N236N Chr.2: 85754897-85754897 on Build GRCh38
345367 c.722C>G p.T241R Chr.2: 85754911-85754911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765012275 Chr.2: 85754924-85754924 on Build GRCh38
rs201179036 Chr.2: 85754855-85754855 on Build GRCh38
rs754089314 Chr.2: 85754997-85754997 on Build GRCh38
rs139460851 Chr.2: 85754859-85754859 on Build GRCh38
rs1021891247 Chr.2: 85754839-85754839 on Build GRCh38
rs772173128 Chr.2: 85754986-85754986 on Build GRCh38
rs780937721 Chr.2: 85754969-85754969 on Build GRCh38
rs748419862 Chr.2: 85754981-85754981 on Build GRCh38
rs777679592 Chr.2: 85754944-85754944 on Build GRCh38
rs766791956 Chr.2: 85754890-85754890 on Build GRCh38
rs767910077 Chr.2: 85754872-85754872 on Build GRCh38
rs546288763 Chr.2: 85754996-85754996 on Build GRCh38
rs774510429 Chr.2: 85754864-85754864 on Build GRCh38
rs553861276 Chr.2: 85754962-85754962 on Build GRCh38
rs747977249 Chr.2: 85754840-85754840 on Build GRCh38
rs76354978 Chr.2: 85754894-85754894 on Build GRCh38
rs764591427 Chr.2: 85754913-85754920 on Build GRCh38
rs746772718 Chr.2: 85754951-85754951 on Build GRCh38
rs374945710 Chr.2: 85754816-85754816 on Build GRCh38
rs749397268 Chr.2: 85754971-85754971 on Build GRCh38
rs768883408 Chr.2: 85754862-85754862 on Build GRCh38
rs774386379 Chr.2: 85754977-85754977 on Build GRCh38
rs755913365 Chr.2: 85754810-85754810 on Build GRCh38
rs1017301315 Chr.2: 85754871-85754871 on Build GRCh38
rs771951468 Chr.2: 85754842-85754842 on Build GRCh38
rs773429076 Chr.2: 85754882-85754882 on Build GRCh38
rs762957319 Chr.2: 85754860-85754860 on Build GRCh38
rs762199518 Chr.2: 85754866-85754866 on Build GRCh38
rs572528543 Chr.2: 85754984-85754984 on Build GRCh38
rs773693435 Chr.2: 85754988-85754988 on Build GRCh38
rs201640710 Chr.2: 85754830-85754830 on Build GRCh38
rs757219438 Chr.2: 85754967-85754967 on Build GRCh38
rs753274495 Chr.2: 85754820-85754820 on Build GRCh38
rs570685156 Chr.2: 85754846-85754846 on Build GRCh38
rs752435998 Chr.2: 85754931-85754931 on Build GRCh38
rs112625718 Chr.2: 85754806-85754806 on Build GRCh38
rs199892675 Chr.2: 85754875-85754875 on Build GRCh38
rs967865255 Chr.2: 85754835-85754835 on Build GRCh38
rs923197079 Chr.2: 85754933-85754933 on Build GRCh38
rs535881836 Chr.2: 85754903-85754903 on Build GRCh38
rs768685045 Chr.2: 85754973-85754973 on Build GRCh38
rs778650050 Chr.2: 85754831-85754831 on Build GRCh38
rs758099463 Chr.2: 85754932-85754932 on Build GRCh38
rs569770902 Chr.2: 85754970-85754970 on Build GRCh38

View Full Product Details