Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 38254802-38255075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC14A
Entrez Gene ID
161198
Gene Name
C-type lectin domain family 14 member A
Gene Aliases
C14orf27, CEG1, EGFR-5
UniGene
Hs.525307
Gene Chromosome Location
Chr.14: 38254000-38256370 on Build GRCh38
Transcripts
NM_175060.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3814780 c.958C>T p.P320S Chr.14: 38255065-38255065 on Build GRCh38
7328202 c.1199T>C p.F400S Chr.14: 38254824-38254824 on Build GRCh38
1369755 c.969C>T p.V323V Chr.14: 38255054-38255054 on Build GRCh38
6015502 c.973G>A p.E325K Chr.14: 38255050-38255050 on Build GRCh38
3690068 c.1220T>C p.V407A Chr.14: 38254803-38254803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375930905 Chr.14: 38255024-38255024 on Build GRCh38
rs765253102 Chr.14: 38254915-38254915 on Build GRCh38
rs750261488 Chr.14: 38254922-38254922 on Build GRCh38
rs138037936 Chr.14: 38254899-38254899 on Build GRCh38
rs148708409 Chr.14: 38255053-38255053 on Build GRCh38
rs778051996 Chr.14: 38254950-38254950 on Build GRCh38
rs76636891 Chr.14: 38254974-38254974 on Build GRCh38
rs201094525 Chr.14: 38254954-38254954 on Build GRCh38
rs779677810 Chr.14: 38254973-38254973 on Build GRCh38
rs369867645 Chr.14: 38254833-38254833 on Build GRCh38
rs757494573 Chr.14: 38254955-38254955 on Build GRCh38
rs780657376 Chr.14: 38255026-38255026 on Build GRCh38
rs1003004217 Chr.14: 38254908-38254908 on Build GRCh38
rs746550618 Chr.14: 38255028-38255028 on Build GRCh38
rs761629295 Chr.14: 38254971-38254971 on Build GRCh38
rs555103437 Chr.14: 38254844-38254844 on Build GRCh38
rs756300764 Chr.14: 38254944-38254944 on Build GRCh38
rs771775423 Chr.14: 38254962-38254962 on Build GRCh38
rs60904056 Chr.14: 38254980-38254980 on Build GRCh38
rs140629728 Chr.14: 38255013-38255013 on Build GRCh38
rs906933537 Chr.14: 38255022-38255022 on Build GRCh38
rs772963880 Chr.14: 38255054-38255054 on Build GRCh38
rs376113989 Chr.14: 38254976-38254976 on Build GRCh38
rs1047159662 Chr.14: 38254859-38254859 on Build GRCh38
rs752987410 Chr.14: 38254936-38254936 on Build GRCh38
rs138633746 Chr.14: 38254871-38254871 on Build GRCh38
rs370513011 Chr.14: 38254998-38254998 on Build GRCh38
rs34744500 Chr.14: 38254920-38254921 on Build GRCh38
rs530380827 Chr.14: 38254836-38254838 on Build GRCh38
rs145445133 Chr.14: 38254895-38254895 on Build GRCh38
rs1012529163 Chr.14: 38254974-38254974 on Build GRCh38
rs370505658 Chr.14: 38255032-38255032 on Build GRCh38
rs751685062 Chr.14: 38254832-38254832 on Build GRCh38
rs757186521 Chr.14: 38254829-38254829 on Build GRCh38
rs1035864538 Chr.14: 38254921-38254921 on Build GRCh38
rs780911738 Chr.14: 38255044-38255044 on Build GRCh38
rs779483782 Chr.14: 38254956-38254956 on Build GRCh38
rs752754237 Chr.14: 38254996-38254996 on Build GRCh38
rs769418112 Chr.14: 38254984-38254984 on Build GRCh38
rs773194890 Chr.14: 38254986-38254986 on Build GRCh38
rs776462742 Chr.14: 38254907-38254907 on Build GRCh38
rs372739405 Chr.14: 38255001-38255001 on Build GRCh38
rs749803064 Chr.14: 38254854-38254854 on Build GRCh38
rs372611166 Chr.14: 38254831-38254831 on Build GRCh38
rs149337036 Chr.14: 38254877-38254877 on Build GRCh38
rs767627256 Chr.14: 38254991-38254991 on Build GRCh38
rs545820191 Chr.14: 38254968-38254968 on Build GRCh38
rs985844647 Chr.14: 38254864-38254864 on Build GRCh38
rs779140207 Chr.14: 38255007-38255007 on Build GRCh38
rs747743367 Chr.14: 38255052-38255052 on Build GRCh38
rs767574364 Chr.14: 38254934-38254934 on Build GRCh38
rs373659027 Chr.14: 38254888-38254888 on Build GRCh38
rs768610750 Chr.14: 38254901-38254901 on Build GRCh38
rs768378999 Chr.14: 38254979-38254979 on Build GRCh38
rs78287081 Chr.14: 38254838-38254838 on Build GRCh38
rs759627401 Chr.14: 38254924-38254924 on Build GRCh38
rs775340249 Chr.14: 38254898-38254898 on Build GRCh38
rs200873816 Chr.14: 38254961-38254961 on Build GRCh38
rs112354022 Chr.14: 38254846-38254846 on Build GRCh38
rs761468083 Chr.14: 38254909-38254909 on Build GRCh38
rs753945066 Chr.14: 38254999-38254999 on Build GRCh38
rs761363362 Chr.14: 38254933-38254934 on Build GRCh38
rs146098376 Chr.14: 38255017-38255017 on Build GRCh38
rs761673996 Chr.14: 38254981-38254981 on Build GRCh38

View Full Product Details