Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108725797-108726038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC7
Entrez Gene ID
163479
Gene Name
fibronectin type III domain containing 7
Gene Aliases
-
UniGene
Hs.258253
Gene Chromosome Location
Chr.1: 108712934-108742749 on Build GRCh38
Transcripts
NM_001144937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4441555 c.1061A>G p.Y354C Chr.1: 108725954-108725954 on Build GRCh38
7099254 c.1035C>A p.F345L Chr.1: 108725928-108725928 on Build GRCh38
3996721 c.1058T>C p.V353A Chr.1: 108725951-108725951 on Build GRCh38
3369175 c.1042T>G p.F348V Chr.1: 108725935-108725935 on Build GRCh38
6862498 c.959_960ins11 p.K321fs*90 Chr.1: 108725852-108725853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761293624 Chr.1: 108725860-108725860 on Build GRCh38
rs146761717 Chr.1: 108725990-108725990 on Build GRCh38
rs374590221 Chr.1: 108725955-108725955 on Build GRCh38
rs780764349 Chr.1: 108725871-108725871 on Build GRCh38
rs754484891 Chr.1: 108725834-108725836 on Build GRCh38
rs771190246 Chr.1: 108725879-108725879 on Build GRCh38
rs754001115 Chr.1: 108725820-108725820 on Build GRCh38
rs765401418 Chr.1: 108725967-108725967 on Build GRCh38
rs376320774 Chr.1: 108725966-108725966 on Build GRCh38
rs748174563 Chr.1: 108725841-108725841 on Build GRCh38
rs748821573 Chr.1: 108726005-108726005 on Build GRCh38
rs774142403 Chr.1: 108725953-108725953 on Build GRCh38
rs752774648 Chr.1: 108725980-108725980 on Build GRCh38
rs755124944 Chr.1: 108725821-108725821 on Build GRCh38
rs761735302 Chr.1: 108725954-108725954 on Build GRCh38
rs575398680 Chr.1: 108725983-108725983 on Build GRCh38
rs749462610 Chr.1: 108725921-108725921 on Build GRCh38
rs778406259 Chr.1: 108725985-108725985 on Build GRCh38
rs558709720 Chr.1: 108725964-108725964 on Build GRCh38
rs199626568 Chr.1: 108725839-108725839 on Build GRCh38
rs192640023 Chr.1: 108725955-108725955 on Build GRCh38
rs373041617 Chr.1: 108725897-108725897 on Build GRCh38
rs370375562 Chr.1: 108725913-108725913 on Build GRCh38
rs1056535710 Chr.1: 108725942-108725942 on Build GRCh38
rs144831878 Chr.1: 108725831-108725831 on Build GRCh38
rs113506602 Chr.1: 108725844-108725844 on Build GRCh38
rs769603053 Chr.1: 108726003-108726003 on Build GRCh38
rs895804995 Chr.1: 108725863-108725863 on Build GRCh38
rs746195690 Chr.1: 108725998-108725998 on Build GRCh38
rs762513456 Chr.1: 108725933-108725933 on Build GRCh38
rs4494160 Chr.1: 108725951-108725951 on Build GRCh38
rs758696908 Chr.1: 108725994-108725994 on Build GRCh38
rs370253483 Chr.1: 108726010-108726010 on Build GRCh38
rs532141533 Chr.1: 108726004-108726004 on Build GRCh38
rs768276314 Chr.1: 108726007-108726007 on Build GRCh38
rs776415530 Chr.1: 108725891-108725891 on Build GRCh38
rs371394123 Chr.1: 108725852-108725852 on Build GRCh38
rs111375246 Chr.1: 108725986-108725986 on Build GRCh38
rs11582005 Chr.1: 108725819-108725819 on Build GRCh38
rs778182588 Chr.1: 108725857-108725857 on Build GRCh38
rs3006870 Chr.1: 108725993-108725993 on Build GRCh38
rs1001233561 Chr.1: 108725996-108725996 on Build GRCh38
rs747832235 Chr.1: 108725907-108725909 on Build GRCh38
rs747410031 Chr.1: 108725873-108725873 on Build GRCh38

View Full Product Details