Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 34042674-34042945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF3
Entrez Gene ID
114899
Gene Name
C1q and tumor necrosis factor related protein 3
Gene Aliases
C1ATNF3, CORCS, CORS, CORS-26, CORS26, CTRP3
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 34017858-34043266 on Build GRCh38
Transcripts
NM_030945.3 NM_181435.5 NR_037951.1

Gene Symbol
C1QTNF3-AMACR
Entrez Gene ID
100534612
Gene Name
C1QTNF3-AMACR readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 33986986-34124528 on Build GRCh38
Transcripts
NM_030945.3 NM_181435.5 NR_037951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3015339 c.254C>T p.P85L Chr.5: 34042872-34042872 on Build GRCh38
6615418 c.255G>A p.P85P Chr.5: 34042871-34042871 on Build GRCh38
243344 c.236T>C p.L79P Chr.5: 34042890-34042890 on Build GRCh38
1578548 c.195_196delTG p.G66fs*4 Chr.5: 34042930-34042931 on Build GRCh38
6512953 c.282G>T p.Q94H Chr.5: 34042844-34042844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369846327 Chr.5: 34042800-34042800 on Build GRCh38
rs984690475 Chr.5: 34042751-34042753 on Build GRCh38
rs1042538186 Chr.5: 34042915-34042915 on Build GRCh38
rs769863322 Chr.5: 34042849-34042849 on Build GRCh38
rs763605813 Chr.5: 34042819-34042819 on Build GRCh38
rs190492922 Chr.5: 34042826-34042826 on Build GRCh38
rs756327553 Chr.5: 34042868-34042868 on Build GRCh38
rs113668504 Chr.5: 34042870-34042870 on Build GRCh38
rs755392798 Chr.5: 34042841-34042841 on Build GRCh38
rs981292320 Chr.5: 34042795-34042795 on Build GRCh38
rs933041044 Chr.5: 34042924-34042924 on Build GRCh38
rs780118939 Chr.5: 34042905-34042905 on Build GRCh38
rs573985404 Chr.5: 34042764-34042765 on Build GRCh38
rs112167551 Chr.5: 34042761-34042762 on Build GRCh38
rs771905676 Chr.5: 34042890-34042890 on Build GRCh38
rs199873478 Chr.5: 34042895-34042895 on Build GRCh38
rs780020256 Chr.5: 34042714-34042714 on Build GRCh38
rs753383673 Chr.5: 34042803-34042803 on Build GRCh38
rs57826552 Chr.5: 34042777-34042778 on Build GRCh38
rs538468468 Chr.5: 34042793-34042794 on Build GRCh38
rs758481865 Chr.5: 34042850-34042850 on Build GRCh38
rs144445678 Chr.5: 34042871-34042871 on Build GRCh38
rs900266305 Chr.5: 34042891-34042891 on Build GRCh38
rs952056918 Chr.5: 34042745-34042745 on Build GRCh38
rs555686985 Chr.5: 34042762-34042764 on Build GRCh38
rs752124283 Chr.5: 34042918-34042918 on Build GRCh38
rs72343197 Chr.5: 34042772-34042773 on Build GRCh38
rs560600894 Chr.5: 34042701-34042701 on Build GRCh38
rs113809897 Chr.5: 34042760-34042760 on Build GRCh38
rs752972223 Chr.5: 34042865-34042865 on Build GRCh38
rs73761686 Chr.5: 34042759-34042759 on Build GRCh38
rs776323791 Chr.5: 34042906-34042906 on Build GRCh38
rs750607069 Chr.5: 34042836-34042838 on Build GRCh38
rs775222105 Chr.5: 34042775-34042775 on Build GRCh38
rs72108923 Chr.5: 34042762-34042763 on Build GRCh38
rs963708650 Chr.5: 34042700-34042700 on Build GRCh38
rs386403521 Chr.5: 34042775-34042776 on Build GRCh38
rs531769358 Chr.5: 34042872-34042872 on Build GRCh38
rs760178805 Chr.5: 34042794-34042794 on Build GRCh38
rs768399020 Chr.5: 34042903-34042903 on Build GRCh38
rs188876397 Chr.5: 34042901-34042901 on Build GRCh38
rs990954680 Chr.5: 34042713-34042713 on Build GRCh38
rs764791785 Chr.5: 34042908-34042908 on Build GRCh38
rs745730855 Chr.5: 34042883-34042883 on Build GRCh38
rs751972062 Chr.5: 34042828-34042828 on Build GRCh38
rs73761687 Chr.5: 34042761-34042761 on Build GRCh38
rs761440829 Chr.5: 34042907-34042907 on Build GRCh38

View Full Product Details