Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7323123-7323348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM4
Entrez Gene ID
341392
Gene Name
acyl-CoA synthetase medium-chain family member 4
Gene Aliases
-
UniGene
Hs.450804
Gene Chromosome Location
Chr.12: 7304332-7328373 on Build GRCh38
Transcripts
NM_001080454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660152 c.1171T>G p.S391A Chr.12: 7323279-7323279 on Build GRCh38
1660154 c.1172C>T p.S391L Chr.12: 7323280-7323280 on Build GRCh38
6553006 c.1169G>T p.G390V Chr.12: 7323277-7323277 on Build GRCh38
943395 c.1184G>A p.G395E Chr.12: 7323292-7323292 on Build GRCh38
695107 c.1126G>A p.G376R Chr.12: 7323234-7323234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053408181 Chr.12: 7323252-7323252 on Build GRCh38
rs774837620 Chr.12: 7323236-7323236 on Build GRCh38
rs778808405 Chr.12: 7323300-7323300 on Build GRCh38
rs758269574 Chr.12: 7323310-7323310 on Build GRCh38
rs978620856 Chr.12: 7323167-7323167 on Build GRCh38
rs1041047159 Chr.12: 7323169-7323169 on Build GRCh38
rs760912619 Chr.12: 7323282-7323282 on Build GRCh38
rs761210824 Chr.12: 7323224-7323224 on Build GRCh38
rs148969232 Chr.12: 7323206-7323206 on Build GRCh38
rs768079091 Chr.12: 7323203-7323203 on Build GRCh38
rs200663706 Chr.12: 7323277-7323277 on Build GRCh38
rs750325732 Chr.12: 7323305-7323305 on Build GRCh38
rs757278847 Chr.12: 7323299-7323299 on Build GRCh38
rs376217473 Chr.12: 7323289-7323289 on Build GRCh38
rs764518698 Chr.12: 7323176-7323176 on Build GRCh38
rs759952698 Chr.12: 7323237-7323237 on Build GRCh38
rs200156270 Chr.12: 7323285-7323285 on Build GRCh38
rs766951990 Chr.12: 7323228-7323228 on Build GRCh38
rs752921586 Chr.12: 7323279-7323279 on Build GRCh38
rs145890307 Chr.12: 7323152-7323152 on Build GRCh38
rs779951447 Chr.12: 7323313-7323313 on Build GRCh38
rs1031013466 Chr.12: 7323291-7323291 on Build GRCh38

View Full Product Details