Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122290953-122291220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD16
Entrez Gene ID
118663
Gene Name
BTB domain containing 16
Gene Aliases
C10orf87
UniGene
Hs.422466
Gene Chromosome Location
Chr.10: 122270745-122338180 on Build GRCh38
Transcripts
NM_001318189.1 NM_144587.3 XM_011539239.2 XM_011539240.2 XM_011539241.2 XM_011539242.2 XM_017015636.1 XM_017015637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1745675 c.586C>T p.Q196* Chr.10: 122291190-122291190 on Build GRCh38
3434996 c.492G>A p.L164L Chr.10: 122291096-122291096 on Build GRCh38
1675623 c.491T>C p.L164P Chr.10: 122291095-122291095 on Build GRCh38
3434997 c.564C>T p.I188I Chr.10: 122291168-122291168 on Build GRCh38
6268294 c.507G>T p.M169I Chr.10: 122291111-122291111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754525137 Chr.10: 122291175-122291176 on Build GRCh38
rs762586523 Chr.10: 122291137-122291137 on Build GRCh38
rs555691014 Chr.10: 122291018-122291018 on Build GRCh38
rs781341697 Chr.10: 122291155-122291155 on Build GRCh38
rs767275051 Chr.10: 122291159-122291159 on Build GRCh38
rs11592545 Chr.10: 122291060-122291060 on Build GRCh38
rs955736780 Chr.10: 122291165-122291165 on Build GRCh38
rs141937724 Chr.10: 122291089-122291089 on Build GRCh38
rs759045854 Chr.10: 122291107-122291107 on Build GRCh38
rs543021361 Chr.10: 122291015-122291015 on Build GRCh38
rs781457098 Chr.10: 122291061-122291061 on Build GRCh38
rs768601482 Chr.10: 122291059-122291059 on Build GRCh38
rs761591973 Chr.10: 122291075-122291075 on Build GRCh38
rs780282713 Chr.10: 122291160-122291160 on Build GRCh38
rs764996066 Chr.10: 122291119-122291119 on Build GRCh38
rs768772900 Chr.10: 122291175-122291175 on Build GRCh38
rs746137609 Chr.10: 122291063-122291063 on Build GRCh38
rs746985229 Chr.10: 122291185-122291185 on Build GRCh38
rs758919125 Chr.10: 122291054-122291054 on Build GRCh38
rs547145447 Chr.10: 122291085-122291085 on Build GRCh38
rs376734976 Chr.10: 122291029-122291029 on Build GRCh38
rs551665654 Chr.10: 122291032-122291032 on Build GRCh38
rs948652501 Chr.10: 122291065-122291065 on Build GRCh38
rs776233499 Chr.10: 122291200-122291200 on Build GRCh38
rs939400398 Chr.10: 122290973-122290973 on Build GRCh38
rs760494805 Chr.10: 122291086-122291086 on Build GRCh38
rs752058130 Chr.10: 122291147-122291147 on Build GRCh38
rs908505195 Chr.10: 122291025-122291025 on Build GRCh38
rs376764537 Chr.10: 122291017-122291017 on Build GRCh38
rs749389215 Chr.10: 122291081-122291081 on Build GRCh38
rs866077792 Chr.10: 122291052-122291052 on Build GRCh38
rs373741777 Chr.10: 122291070-122291070 on Build GRCh38
rs764669031 Chr.10: 122291138-122291138 on Build GRCh38
rs369288423 Chr.10: 122291148-122291148 on Build GRCh38
rs892780723 Chr.10: 122291091-122291091 on Build GRCh38
rs150125169 Chr.10: 122291188-122291188 on Build GRCh38
rs997091099 Chr.10: 122291156-122291156 on Build GRCh38
rs983273774 Chr.10: 122291021-122291021 on Build GRCh38
rs764910080 Chr.10: 122291034-122291034 on Build GRCh38
rs753674597 Chr.10: 122291174-122291175 on Build GRCh38
rs149699378 Chr.10: 122291133-122291133 on Build GRCh38
rs976462540 Chr.10: 122291113-122291113 on Build GRCh38
rs140767498 Chr.10: 122291076-122291076 on Build GRCh38
rs1011176667 Chr.10: 122291101-122291102 on Build GRCh38
rs776344285 Chr.10: 122291090-122291090 on Build GRCh38
rs752261857 Chr.10: 122291036-122291036 on Build GRCh38
rs928463457 Chr.10: 122291084-122291084 on Build GRCh38
rs749334573 Chr.10: 122291172-122291172 on Build GRCh38

View Full Product Details