Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116403795-116404064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A1
Entrez Gene ID
476
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Gene Aliases
-
UniGene
Hs.371889
Gene Chromosome Location
Chr.1: 116373173-116404774 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027645.1 NR_027646.1

Gene Symbol
ATP1A1-AS1
Entrez Gene ID
84852
Gene Name
ATP1A1 antisense RNA 1
Gene Aliases
ATP1A1OS, C1orf203
UniGene
Hs.664752
Gene Chromosome Location
Chr.1: 116392865-116418622 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1 NM_001160234.1 XM_017001360.1 XM_017001361.1 NR_027645.1 NR_027646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4760474 c.3035G>A p.R1012H Chr.1: 116403967-116403967 on Build GRCh38
6356390 c.2963G>A p.W988* Chr.1: 116403895-116403895 on Build GRCh38
894400 c.2999T>C p.V1000A Chr.1: 116403931-116403931 on Build GRCh38
1333027 c.2990_2992delTCA p.I998delI Chr.1: 116403922-116403924 on Build GRCh38
309354 c.3031C>T p.R1011* Chr.1: 116403963-116403963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932219449 Chr.1: 116404029-116404029 on Build GRCh38
rs773244746 Chr.1: 116404021-116404021 on Build GRCh38
rs12139437 Chr.1: 116403983-116403983 on Build GRCh38
rs146071470 Chr.1: 116403938-116403938 on Build GRCh38
rs369244764 Chr.1: 116403833-116403833 on Build GRCh38
rs995342661 Chr.1: 116403819-116403819 on Build GRCh38
rs769256751 Chr.1: 116403999-116403999 on Build GRCh38
rs770455038 Chr.1: 116403857-116403857 on Build GRCh38
rs76876899 Chr.1: 116404036-116404036 on Build GRCh38
rs529546474 Chr.1: 116404033-116404033 on Build GRCh38
rs546749593 Chr.1: 116403853-116403854 on Build GRCh38
rs777390048 Chr.1: 116403849-116403849 on Build GRCh38
rs199565723 Chr.1: 116404003-116404003 on Build GRCh38
rs371751183 Chr.1: 116403834-116403834 on Build GRCh38
rs752427437 Chr.1: 116404003-116404005 on Build GRCh38
rs970827565 Chr.1: 116403887-116403887 on Build GRCh38
rs10924083 Chr.1: 116404013-116404013 on Build GRCh38
rs748414419 Chr.1: 116404006-116404006 on Build GRCh38
rs749878372 Chr.1: 116403998-116403998 on Build GRCh38
rs575290482 Chr.1: 116403870-116403870 on Build GRCh38
rs12136995 Chr.1: 116403847-116403847 on Build GRCh38
rs762137894 Chr.1: 116403877-116403877 on Build GRCh38
rs752411608 Chr.1: 116403950-116403950 on Build GRCh38
rs746455839 Chr.1: 116403850-116403850 on Build GRCh38
rs774576756 Chr.1: 116403872-116403872 on Build GRCh38
rs771215472 Chr.1: 116403889-116403889 on Build GRCh38
rs535384986 Chr.1: 116403848-116403848 on Build GRCh38
rs780836962 Chr.1: 116403990-116403990 on Build GRCh38
rs758090295 Chr.1: 116403964-116403964 on Build GRCh38
rs987713450 Chr.1: 116404032-116404032 on Build GRCh38
rs374509870 Chr.1: 116403929-116403929 on Build GRCh38
rs773368360 Chr.1: 116403905-116403905 on Build GRCh38
rs746521823 Chr.1: 116403967-116403967 on Build GRCh38
rs545614799 Chr.1: 116403890-116403890 on Build GRCh38
rs777250534 Chr.1: 116403966-116403966 on Build GRCh38
rs760709044 Chr.1: 116403908-116403908 on Build GRCh38
rs533448995 Chr.1: 116403824-116403827 on Build GRCh38
rs977860799 Chr.1: 116403920-116403920 on Build GRCh38
rs754902079 Chr.1: 116403932-116403932 on Build GRCh38
rs368714905 Chr.1: 116403868-116403868 on Build GRCh38
rs776074207 Chr.1: 116403866-116403866 on Build GRCh38
rs1024623627 Chr.1: 116403874-116403874 on Build GRCh38
rs753827120 Chr.1: 116403930-116403930 on Build GRCh38

View Full Product Details