Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88728456-88728729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13A
Entrez Gene ID
10144
Gene Name
family with sequence similarity 13 member A
Gene Aliases
ARHGAP48, FAM13A1
UniGene
Hs.714536
Gene Chromosome Location
Chr.4: 88725954-89111398 on Build GRCh38
Transcripts
NM_001015045.2 NM_001265578.1 NM_001265579.1 NM_001265580.1 NM_014883.3 XM_005262681.2 XM_005262682.2 XM_005262683.2 XM_005262684.3 XM_006714057.3 XM_011531516.1 XM_011531517.1 XM_011531518.1 XM_011531519.2 XM_017007624.1 XM_017007625.1 XM_017007626.1 XM_017007627.1 XM_017007628.1 XM_017007629.1 XM_017007630.1 XM_017007631.1 XM_017007632.1 XM_017007633.1 XM_017007634.1 NR_002806.2

Gene Symbol
FAM13A-AS1
Entrez Gene ID
285512
Gene Name
FAM13A antisense RNA 1
Gene Aliases
FAM13A1OS, FAM13AOS, NCRNA00039
UniGene
Hs.744957
Gene Chromosome Location
Chr.4: 88709789-88730103 on Build GRCh38
Transcripts
NM_001015045.2 NM_001265578.1 NM_001265579.1 NM_001265580.1 NM_014883.3 XM_005262681.2 XM_005262682.2 XM_005262683.2 XM_005262684.3 XM_006714057.3 XM_011531516.1 XM_011531517.1 XM_011531518.1 XM_011531519.2 XM_017007624.1 XM_017007625.1 XM_017007626.1 XM_017007627.1 XM_017007628.1 XM_017007629.1 XM_017007630.1 XM_017007631.1 XM_017007632.1 XM_017007633.1 XM_017007634.1 NR_002806.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
481677 c.3031G>A p.V1011M Chr.4: 88728574-88728574 on Build GRCh38
7423098 c.2029A>C p.K677Q Chr.4: 88728598-88728598 on Build GRCh38
6168330 c.2085G>C p.K695N Chr.4: 88728542-88728542 on Build GRCh38
481676 c.3032T>A p.V1011E Chr.4: 88728573-88728573 on Build GRCh38
6250013 c.3011C>A p.A1004E Chr.4: 88728594-88728594 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760453733 Chr.4: 88728631-88728631 on Build GRCh38
rs562487263 Chr.4: 88728530-88728530 on Build GRCh38
rs773057345 Chr.4: 88728549-88728549 on Build GRCh38
rs955259348 Chr.4: 88728602-88728602 on Build GRCh38
rs964010449 Chr.4: 88728702-88728702 on Build GRCh38
rs747797735 Chr.4: 88728701-88728701 on Build GRCh38
rs759231694 Chr.4: 88728639-88728639 on Build GRCh38
rs767370996 Chr.4: 88728485-88728485 on Build GRCh38
rs186862605 Chr.4: 88728560-88728560 on Build GRCh38
rs573977255 Chr.4: 88728510-88728510 on Build GRCh38
rs368002011 Chr.4: 88728518-88728518 on Build GRCh38
rs766119027 Chr.4: 88728637-88728637 on Build GRCh38
rs1001624447 Chr.4: 88728541-88728541 on Build GRCh38
rs376624008 Chr.4: 88728686-88728686 on Build GRCh38
rs73841658 Chr.4: 88728525-88728525 on Build GRCh38
rs182426402 Chr.4: 88728493-88728493 on Build GRCh38
rs149073720 Chr.4: 88728559-88728559 on Build GRCh38
rs538935702 Chr.4: 88728555-88728555 on Build GRCh38
rs768919981 Chr.4: 88728583-88728583 on Build GRCh38
rs770041835 Chr.4: 88728517-88728517 on Build GRCh38
rs371740753 Chr.4: 88728656-88728656 on Build GRCh38
rs920774354 Chr.4: 88728478-88728478 on Build GRCh38
rs749295751 Chr.4: 88728528-88728528 on Build GRCh38
rs764659890 Chr.4: 88728481-88728481 on Build GRCh38
rs767274249 Chr.4: 88728594-88728594 on Build GRCh38
rs767397872 Chr.4: 88728532-88728532 on Build GRCh38
rs754551092 Chr.4: 88728499-88728499 on Build GRCh38
rs748359167 Chr.4: 88728555-88728556 on Build GRCh38
rs746183360 Chr.4: 88728569-88728569 on Build GRCh38
rs776617204 Chr.4: 88728638-88728638 on Build GRCh38
rs747540136 Chr.4: 88728507-88728507 on Build GRCh38
rs546892366 Chr.4: 88728593-88728593 on Build GRCh38
rs772036495 Chr.4: 88728609-88728609 on Build GRCh38
rs780205766 Chr.4: 88728579-88728579 on Build GRCh38
rs761720527 Chr.4: 88728531-88728531 on Build GRCh38
rs781605328 Chr.4: 88728568-88728568 on Build GRCh38
rs752294802 Chr.4: 88728563-88728563 on Build GRCh38
rs778357921 Chr.4: 88728503-88728503 on Build GRCh38
rs773038490 Chr.4: 88728611-88728611 on Build GRCh38
rs141659205 Chr.4: 88728498-88728498 on Build GRCh38
rs756437230 Chr.4: 88728570-88728570 on Build GRCh38
rs756447316 Chr.4: 88728693-88728693 on Build GRCh38
rs116035805 Chr.4: 88728564-88728564 on Build GRCh38
rs767998593 Chr.4: 88728666-88728666 on Build GRCh38
rs757857060 Chr.4: 88728657-88728657 on Build GRCh38
rs532910718 Chr.4: 88728558-88728558 on Build GRCh38
rs143085078 Chr.4: 88728581-88728581 on Build GRCh38
rs765047420 Chr.4: 88728653-88728653 on Build GRCh38
rs774386529 Chr.4: 88728585-88728585 on Build GRCh38
rs760446820 Chr.4: 88728552-88728552 on Build GRCh38
rs1011474529 Chr.4: 88728494-88728494 on Build GRCh38
rs750237201 Chr.4: 88728486-88728486 on Build GRCh38
rs746282559 Chr.4: 88728516-88728516 on Build GRCh38

View Full Product Details