Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 158714010-158714283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBF1
Entrez Gene ID
1879
Gene Name
early B-cell factor 1
Gene Aliases
COE1, EBF, O/E-1, OLF1
UniGene
Hs.573143
Gene Chromosome Location
Chr.5: 158695915-159099762 on Build GRCh38
Transcripts
NM_001290360.2 NM_001324101.1 NM_001324103.1 NM_001324106.1 NM_001324107.1 NM_001324108.1 NM_001324109.1 NM_001324111.1 NM_024007.4 NM_182708.2 XM_017009192.1 XM_017009193.1 XM_017009194.1 XM_017009195.1 XM_017009196.1 XM_017009197.1 XM_017009198.1 XM_017009199.1 XM_017009200.1 XM_017009201.1 XM_017009204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651124 c.1153C>A p.L385M Chr.5: 158714155-158714155 on Build GRCh38
3613364 c.1135C>T p.L379F Chr.5: 158714173-158714173 on Build GRCh38
1065107 c.1129G>T p.V377L Chr.5: 158714179-158714179 on Build GRCh38
736994 c.1142G>T p.R381M Chr.5: 158714166-158714166 on Build GRCh38
736993 c.1126G>T p.E376* Chr.5: 158714182-158714182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758428545 Chr.5: 158714124-158714124 on Build GRCh38
rs1022361914 Chr.5: 158714243-158714243 on Build GRCh38
rs774806946 Chr.5: 158714076-158714076 on Build GRCh38
rs766830246 Chr.5: 158714067-158714067 on Build GRCh38
rs781066735 Chr.5: 158714129-158714129 on Build GRCh38
rs763330092 Chr.5: 158714066-158714066 on Build GRCh38
rs773452840 Chr.5: 158714153-158714153 on Build GRCh38
rs757090060 Chr.5: 158714111-158714111 on Build GRCh38
rs931717615 Chr.5: 158714073-158714073 on Build GRCh38
rs771261200 Chr.5: 158714159-158714159 on Build GRCh38
rs1017600494 Chr.5: 158714050-158714050 on Build GRCh38
rs879180386 Chr.5: 158714049-158714049 on Build GRCh38
rs760087607 Chr.5: 158714210-158714211 on Build GRCh38
rs990852982 Chr.5: 158714241-158714241 on Build GRCh38
rs760267863 Chr.5: 158714231-158714231 on Build GRCh38
rs752316005 Chr.5: 158714082-158714082 on Build GRCh38
rs151118528 Chr.5: 158714110-158714110 on Build GRCh38
rs769858589 Chr.5: 158714150-158714150 on Build GRCh38
rs768070173 Chr.5: 158714179-158714179 on Build GRCh38
rs917163757 Chr.5: 158714226-158714226 on Build GRCh38
rs763834569 Chr.5: 158714108-158714108 on Build GRCh38
rs887589618 Chr.5: 158714079-158714079 on Build GRCh38
rs968557815 Chr.5: 158714263-158714263 on Build GRCh38
rs577902209 Chr.5: 158714043-158714043 on Build GRCh38
rs900172183 Chr.5: 158714044-158714044 on Build GRCh38
rs745789186 Chr.5: 158714121-158714121 on Build GRCh38
rs199852631 Chr.5: 158714078-158714078 on Build GRCh38
rs749550787 Chr.5: 158714158-158714158 on Build GRCh38
rs748273958 Chr.5: 158714141-158714141 on Build GRCh38
rs774753560 Chr.5: 158714161-158714161 on Build GRCh38
rs778903431 Chr.5: 158714114-158714114 on Build GRCh38
rs755651580 Chr.5: 158714107-158714107 on Build GRCh38
rs370368323 Chr.5: 158714222-158714222 on Build GRCh38
rs759953915 Chr.5: 158714164-158714164 on Build GRCh38
rs959397532 Chr.5: 158714225-158714225 on Build GRCh38
rs574104675 Chr.5: 158714244-158714244 on Build GRCh38

View Full Product Details