Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74338462-74338709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11A1
Entrez Gene ID
1583
Gene Name
cytochrome P450 family 11 subfamily A member 1
Gene Aliases
CYP11A, CYPXIA1, P450SCC
UniGene
Hs.303980
Gene Chromosome Location
Chr.15: 74337762-74367740 on Build GRCh38
Transcripts
NM_000781.2 NM_001099773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5849763 c.1411G>A p.E471K Chr.15: 74338594-74338594 on Build GRCh38
3887488 c.1351C>T p.R451W Chr.15: 74338654-74338654 on Build GRCh38
271189 c.1401C>T p.I467I Chr.15: 74338604-74338604 on Build GRCh38
6638996 c.1380G>A p.R460R Chr.15: 74338625-74338625 on Build GRCh38
434288 c.1341C>T p.I447I Chr.15: 74338664-74338664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566367949 Chr.15: 74338493-74338493 on Build GRCh38
rs763985808 Chr.15: 74338597-74338597 on Build GRCh38
rs749515942 Chr.15: 74338616-74338616 on Build GRCh38
rs980016533 Chr.15: 74338637-74338637 on Build GRCh38
rs146830942 Chr.15: 74338670-74338670 on Build GRCh38
rs141235847 Chr.15: 74338612-74338612 on Build GRCh38
rs372523428 Chr.15: 74338610-74338610 on Build GRCh38
rs748510040 Chr.15: 74338618-74338618 on Build GRCh38
rs961597307 Chr.15: 74338482-74338482 on Build GRCh38
rs773746289 Chr.15: 74338540-74338540 on Build GRCh38
rs376136531 Chr.15: 74338531-74338531 on Build GRCh38
rs757055824 Chr.15: 74338603-74338603 on Build GRCh38
rs781028010 Chr.15: 74338604-74338604 on Build GRCh38
rs914444285 Chr.15: 74338580-74338580 on Build GRCh38
rs918002510 Chr.15: 74338608-74338608 on Build GRCh38
rs759185471 Chr.15: 74338555-74338555 on Build GRCh38
rs761206886 Chr.15: 74338650-74338650 on Build GRCh38
rs567242447 Chr.15: 74338530-74338530 on Build GRCh38
rs368047178 Chr.15: 74338609-74338609 on Build GRCh38
rs1019309635 Chr.15: 74338552-74338552 on Build GRCh38
rs774799229 Chr.15: 74338617-74338617 on Build GRCh38
rs1054131477 Chr.15: 74338678-74338678 on Build GRCh38
rs953733004 Chr.15: 74338520-74338521 on Build GRCh38
rs749547595 Chr.15: 74338673-74338673 on Build GRCh38
rs774511568 Chr.15: 74338520-74338520 on Build GRCh38
rs535782968 Chr.15: 74338627-74338627 on Build GRCh38
rs538118268 Chr.15: 74338659-74338659 on Build GRCh38
rs778230506 Chr.15: 74338662-74338662 on Build GRCh38
rs750370186 Chr.15: 74338682-74338682 on Build GRCh38
rs149795879 Chr.15: 74338502-74338502 on Build GRCh38
rs767327902 Chr.15: 74338680-74338680 on Build GRCh38
rs773652136 Chr.15: 74338644-74338644 on Build GRCh38
rs775102947 Chr.15: 74338654-74338654 on Build GRCh38
rs764826737 Chr.15: 74338557-74338557 on Build GRCh38
rs185821614 Chr.15: 74338489-74338489 on Build GRCh38
rs764844923 Chr.15: 74338653-74338653 on Build GRCh38
rs752413112 Chr.15: 74338583-74338583 on Build GRCh38
rs527463931 Chr.15: 74338498-74338498 on Build GRCh38
rs758061011 Chr.15: 74338591-74338591 on Build GRCh38

View Full Product Details