Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1225353-1225584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBP1
Entrez Gene ID
1487
Gene Name
C-terminal binding protein 1
Gene Aliases
BARS
UniGene
Hs.208597
Gene Chromosome Location
Chr.4: 1211440-1250329 on Build GRCh38
Transcripts
NM_001012614.1 NM_001328.2 XM_005272261.3 XM_005272263.4 XM_011513395.2 XM_011513396.2 XM_011513397.2 XM_017007762.1 XM_017007763.1 XM_017007764.1 XM_017007765.1 XM_017007766.1 XM_017007767.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7245876 c.380A>C p.E127A Chr.4: 1225527-1225527 on Build GRCh38
6045053 c.478G>A p.E160K Chr.4: 1225429-1225429 on Build GRCh38
7276407 c.446C>T p.A149V Chr.4: 1225461-1225461 on Build GRCh38
5038538 c.455A>G p.E152G Chr.4: 1225452-1225452 on Build GRCh38
5636727 c.421C>T p.R141W Chr.4: 1225486-1225486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773892800 Chr.4: 1225403-1225403 on Build GRCh38
rs1048261948 Chr.4: 1225481-1225481 on Build GRCh38
rs375752732 Chr.4: 1225425-1225425 on Build GRCh38
rs111349558 Chr.4: 1225544-1225544 on Build GRCh38
rs778804693 Chr.4: 1225514-1225514 on Build GRCh38
rs556334680 Chr.4: 1225387-1225387 on Build GRCh38
rs1039008281 Chr.4: 1225490-1225490 on Build GRCh38
rs1020093537 Chr.4: 1225524-1225524 on Build GRCh38
rs1047769350 Chr.4: 1225541-1225541 on Build GRCh38
rs139564947 Chr.4: 1225430-1225430 on Build GRCh38
rs889585115 Chr.4: 1225493-1225493 on Build GRCh38
rs771867667 Chr.4: 1225520-1225520 on Build GRCh38
rs765831710 Chr.4: 1225391-1225391 on Build GRCh38
rs866608802 Chr.4: 1225376-1225376 on Build GRCh38
rs370399291 Chr.4: 1225460-1225460 on Build GRCh38
rs745447562 Chr.4: 1225519-1225519 on Build GRCh38
rs1045458 Chr.4: 1225388-1225388 on Build GRCh38
rs868779152 Chr.4: 1225417-1225417 on Build GRCh38
rs372503163 Chr.4: 1225409-1225409 on Build GRCh38
rs535413566 Chr.4: 1225405-1225405 on Build GRCh38
rs756957912 Chr.4: 1225461-1225461 on Build GRCh38
rs749298564 Chr.4: 1225447-1225447 on Build GRCh38
rs989650064 Chr.4: 1225559-1225559 on Build GRCh38
rs756383970 Chr.4: 1225550-1225550 on Build GRCh38
rs773539612 Chr.4: 1225549-1225549 on Build GRCh38
rs965475865 Chr.4: 1225538-1225538 on Build GRCh38
rs775346532 Chr.4: 1225523-1225523 on Build GRCh38
rs988524723 Chr.4: 1225379-1225379 on Build GRCh38
rs866127430 Chr.4: 1225427-1225427 on Build GRCh38
rs746485008 Chr.4: 1225543-1225543 on Build GRCh38
rs955465249 Chr.4: 1225452-1225452 on Build GRCh38
rs777656363 Chr.4: 1225432-1225432 on Build GRCh38
rs553745756 Chr.4: 1225406-1225406 on Build GRCh38
rs573418618 Chr.4: 1225418-1225418 on Build GRCh38

View Full Product Details