Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11444196-11444429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM167A
Entrez Gene ID
83648
Gene Name
family with sequence similarity 167 member A
Gene Aliases
C8orf13, D8S265
UniGene
Hs.124299
Gene Chromosome Location
Chr.8: 11421464-11475904 on Build GRCh38
Transcripts
NM_053279.2 XM_005272398.4 XM_011543837.1 XM_011543838.2 XM_011543840.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5044383 c.47G>A p.G16E Chr.8: 11444365-11444365 on Build GRCh38
5062813 c.183G>A p.P61P Chr.8: 11444229-11444229 on Build GRCh38
7093504 c.198G>A p.E66E Chr.8: 11444214-11444214 on Build GRCh38
7207523 c.199C>T p.P67S Chr.8: 11444213-11444213 on Build GRCh38
453874 c.155G>T p.R52M Chr.8: 11444257-11444257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149650089 Chr.8: 11444336-11444336 on Build GRCh38
rs759625004 Chr.8: 11444272-11444272 on Build GRCh38
rs770193242 Chr.8: 11444383-11444383 on Build GRCh38
rs770700939 Chr.8: 11444229-11444229 on Build GRCh38
rs373958877 Chr.8: 11444309-11444309 on Build GRCh38
rs775338153 Chr.8: 11444331-11444331 on Build GRCh38
rs762521323 Chr.8: 11444234-11444234 on Build GRCh38
rs768290619 Chr.8: 11444361-11444361 on Build GRCh38
rs772753147 Chr.8: 11444235-11444235 on Build GRCh38
rs746858234 Chr.8: 11444226-11444226 on Build GRCh38
rs771345709 Chr.8: 11444325-11444325 on Build GRCh38
rs774376018 Chr.8: 11444401-11444401 on Build GRCh38
rs771384390 Chr.8: 11444369-11444369 on Build GRCh38
rs148539546 Chr.8: 11444373-11444373 on Build GRCh38
rs199614470 Chr.8: 11444352-11444352 on Build GRCh38
rs375358571 Chr.8: 11444230-11444230 on Build GRCh38
rs374097546 Chr.8: 11444329-11444329 on Build GRCh38
rs767856136 Chr.8: 11444402-11444402 on Build GRCh38
rs755086250 Chr.8: 11444217-11444217 on Build GRCh38
rs758295577 Chr.8: 11444222-11444222 on Build GRCh38
rs147319341 Chr.8: 11444224-11444224 on Build GRCh38
rs960661017 Chr.8: 11444404-11444404 on Build GRCh38
rs370343498 Chr.8: 11444318-11444318 on Build GRCh38
rs367989433 Chr.8: 11444351-11444351 on Build GRCh38
rs969002888 Chr.8: 11444220-11444220 on Build GRCh38
rs767410260 Chr.8: 11444410-11444410 on Build GRCh38
rs894496900 Chr.8: 11444354-11444354 on Build GRCh38
rs781152754 Chr.8: 11444349-11444349 on Build GRCh38
rs745557831 Chr.8: 11444308-11444308 on Build GRCh38
rs757052248 Chr.8: 11444302-11444302 on Build GRCh38
rs899441070 Chr.8: 11444282-11444282 on Build GRCh38
rs774822079 Chr.8: 11444231-11444231 on Build GRCh38
rs940870500 Chr.8: 11444251-11444251 on Build GRCh38
rs908002984 Chr.8: 11444248-11444248 on Build GRCh38
rs140103747 Chr.8: 11444343-11444343 on Build GRCh38
rs574660509 Chr.8: 11444288-11444288 on Build GRCh38
rs559475589 Chr.8: 11444283-11444283 on Build GRCh38
rs765394039 Chr.8: 11444277-11444277 on Build GRCh38
rs753999118 Chr.8: 11444216-11444216 on Build GRCh38
rs368969734 Chr.8: 11444377-11444377 on Build GRCh38
rs3021513 Chr.8: 11444244-11444244 on Build GRCh38
rs754100470 Chr.8: 11444259-11444259 on Build GRCh38
rs146121515 Chr.8: 11444342-11444342 on Build GRCh38
rs752763578 Chr.8: 11444284-11444284 on Build GRCh38
rs769370993 Chr.8: 11444310-11444310 on Build GRCh38
rs370977636 Chr.8: 11444362-11444362 on Build GRCh38
rs376264572 Chr.8: 11444242-11444242 on Build GRCh38
rs748864218 Chr.8: 11444317-11444317 on Build GRCh38
rs199694475 Chr.8: 11444301-11444301 on Build GRCh38
rs565901675 Chr.8: 11444409-11444409 on Build GRCh38
rs775694535 Chr.8: 11444390-11444390 on Build GRCh38
rs35953795 Chr.8: 11444324-11444324 on Build GRCh38
rs563618320 Chr.8: 11444303-11444303 on Build GRCh38
rs143805495 Chr.8: 11444348-11444348 on Build GRCh38
rs201387976 Chr.8: 11444330-11444330 on Build GRCh38
rs146805791 Chr.8: 11444356-11444356 on Build GRCh38
rs375488017 Chr.8: 11444391-11444391 on Build GRCh38
rs761201168 Chr.8: 11444324-11444324 on Build GRCh38
rs573857490 Chr.8: 11444392-11444392 on Build GRCh38
rs777749973 Chr.8: 11444291-11444291 on Build GRCh38
rs370162158 Chr.8: 11444218-11444218 on Build GRCh38

View Full Product Details