Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75481771-75482040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIPR1
Entrez Gene ID
11010
Gene Name
GLI pathogenesis related 1
Gene Aliases
CRISP7, GLIPR, RTVP1
UniGene
Hs.205558
Gene Chromosome Location
Chr.12: 75480733-75501936 on Build GRCh38
Transcripts
NM_006851.2 XM_011537775.2 XM_017018721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2102490 c.187G>A p.A63T Chr.12: 75481846-75481846 on Build GRCh38
50237 c.259_260insC p.H87fs*33 Chr.12: 75481918-75481919 on Build GRCh38
4940311 c.327T>A p.S109S Chr.12: 75481986-75481986 on Build GRCh38
325403 c.248T>A p.L83Q Chr.12: 75481907-75481907 on Build GRCh38
5531728 c.326C>T p.S109F Chr.12: 75481985-75481985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778700482 Chr.12: 75481897-75481897 on Build GRCh38
rs758168594 Chr.12: 75482010-75482010 on Build GRCh38
rs752946024 Chr.12: 75481886-75481886 on Build GRCh38
rs771458888 Chr.12: 75481965-75481965 on Build GRCh38
rs937761950 Chr.12: 75481808-75481808 on Build GRCh38
rs779492735 Chr.12: 75481821-75481821 on Build GRCh38
rs200477933 Chr.12: 75481926-75481926 on Build GRCh38
rs778628400 Chr.12: 75481809-75481809 on Build GRCh38
rs781595510 Chr.12: 75481803-75481803 on Build GRCh38
rs201158358 Chr.12: 75481816-75481816 on Build GRCh38
rs543406828 Chr.12: 75481953-75481953 on Build GRCh38
rs140910647 Chr.12: 75481884-75481884 on Build GRCh38
rs751700531 Chr.12: 75481795-75481795 on Build GRCh38
rs754131332 Chr.12: 75482005-75482005 on Build GRCh38
rs779938880 Chr.12: 75481913-75481913 on Build GRCh38
rs375183295 Chr.12: 75481804-75481804 on Build GRCh38
rs199803624 Chr.12: 75481849-75481849 on Build GRCh38
rs757050946 Chr.12: 75481807-75481807 on Build GRCh38
rs1020218152 Chr.12: 75481989-75481989 on Build GRCh38
rs141837973 Chr.12: 75481930-75481930 on Build GRCh38
rs767699589 Chr.12: 75481994-75481994 on Build GRCh38
rs148893348 Chr.12: 75481918-75481918 on Build GRCh38
rs760892233 Chr.12: 75481996-75481996 on Build GRCh38
rs769346384 Chr.12: 75481943-75481943 on Build GRCh38
rs954119520 Chr.12: 75481955-75481955 on Build GRCh38
rs143594819 Chr.12: 75481923-75481923 on Build GRCh38
rs764255439 Chr.12: 75482001-75482001 on Build GRCh38
rs116107307 Chr.12: 75481929-75481929 on Build GRCh38
rs751649138 Chr.12: 75481881-75481881 on Build GRCh38
rs369269907 Chr.12: 75481854-75481854 on Build GRCh38
rs201262620 Chr.12: 75481843-75481843 on Build GRCh38
rs745811065 Chr.12: 75481814-75481814 on Build GRCh38
rs377252118 Chr.12: 75481951-75481951 on Build GRCh38
rs768145196 Chr.12: 75481924-75481924 on Build GRCh38
rs774972153 Chr.12: 75481968-75481968 on Build GRCh38
rs138690767 Chr.12: 75481903-75481903 on Build GRCh38
rs373244879 Chr.12: 75481986-75481986 on Build GRCh38
rs373512529 Chr.12: 75481831-75481831 on Build GRCh38
rs752722580 Chr.12: 75481995-75481995 on Build GRCh38
rs767014464 Chr.12: 75481861-75481861 on Build GRCh38
rs202037173 Chr.12: 75481859-75481859 on Build GRCh38
rs141682565 Chr.12: 75481887-75481887 on Build GRCh38
rs1909979 Chr.12: 75481796-75481796 on Build GRCh38
rs1054914882 Chr.12: 75481813-75481813 on Build GRCh38
rs1021147115 Chr.12: 75481879-75481879 on Build GRCh38
rs377664018 Chr.12: 75482013-75482013 on Build GRCh38
rs763254899 Chr.12: 75481957-75481957 on Build GRCh38
rs1056505225 Chr.12: 75481908-75481908 on Build GRCh38
rs150528235 Chr.12: 75481904-75481904 on Build GRCh38
rs773455551 Chr.12: 75481855-75481855 on Build GRCh38
rs985941995 Chr.12: 75482012-75482012 on Build GRCh38
rs759656765 Chr.12: 75481882-75481882 on Build GRCh38

View Full Product Details