Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 91962800-91963049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRDT
Entrez Gene ID
676
Gene Name
bromodomain testis associated
Gene Aliases
BRD6, CT9
UniGene
Hs.482520
Gene Chromosome Location
Chr.1: 91949371-92014428 on Build GRCh38
Transcripts
NM_001242805.2 NM_001242806.2 NM_001242807.2 NM_001242808.2 NM_001242810.2 NM_001726.4 NM_207189.3 XM_006710853.3 XM_006710854.3 XM_006710855.3 XM_006710856.3 XM_006710857.3 XM_011542032.2 XM_011542033.2 XM_011542034.2 XM_011542035.2 XM_011542036.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6501176 c.180A>T p.K60N Chr.1: 91962934-91962934 on Build GRCh38
6507500 c.82T>A p.L28M Chr.1: 91962836-91962836 on Build GRCh38
3866239 c.94C>A p.L32I Chr.1: 91962848-91962848 on Build GRCh38
6305963 c.73A>T p.N25Y Chr.1: 91962827-91962827 on Build GRCh38
4889105 c.116T>C p.V39A Chr.1: 91962870-91962870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759909807 Chr.1: 91962926-91962926 on Build GRCh38
rs753146796 Chr.1: 91962844-91962844 on Build GRCh38
rs779401099 Chr.1: 91962843-91962843 on Build GRCh38
rs779804352 Chr.1: 91962970-91962970 on Build GRCh38
rs149333024 Chr.1: 91962931-91962931 on Build GRCh38
rs780711100 Chr.1: 91962968-91962968 on Build GRCh38
rs768759964 Chr.1: 91962993-91962993 on Build GRCh38
rs748249353 Chr.1: 91962976-91962976 on Build GRCh38
rs754180862 Chr.1: 91962838-91962838 on Build GRCh38
rs780848368 Chr.1: 91962872-91962872 on Build GRCh38
rs751053540 Chr.1: 91962951-91962951 on Build GRCh38
rs775579567 Chr.1: 91962888-91962888 on Build GRCh38
rs745680057 Chr.1: 91962878-91962878 on Build GRCh38
rs749056697 Chr.1: 91962990-91962990 on Build GRCh38
rs758944987 Chr.1: 91962940-91962940 on Build GRCh38
rs755813321 Chr.1: 91962971-91962971 on Build GRCh38
rs371179548 Chr.1: 91962914-91962914 on Build GRCh38
rs189969715 Chr.1: 91962857-91962857 on Build GRCh38
rs749293993 Chr.1: 91962889-91962889 on Build GRCh38
rs780058106 Chr.1: 91962978-91962978 on Build GRCh38
rs776849012 Chr.1: 91962923-91962923 on Build GRCh38
rs376824899 Chr.1: 91962851-91962851 on Build GRCh38
rs138654452 Chr.1: 91962901-91962901 on Build GRCh38
rs768741918 Chr.1: 91962956-91962956 on Build GRCh38
rs746497939 Chr.1: 91962855-91962855 on Build GRCh38
rs371597005 Chr.1: 91962915-91962915 on Build GRCh38
rs10783071 Chr.1: 91962938-91962938 on Build GRCh38
rs769643899 Chr.1: 91962886-91962886 on Build GRCh38
rs868740528 Chr.1: 91962852-91962852 on Build GRCh38
rs950933060 Chr.1: 91962867-91962867 on Build GRCh38
rs769747935 Chr.1: 91962981-91962983 on Build GRCh38
rs138274836 Chr.1: 91962847-91962847 on Build GRCh38
rs201337329 Chr.1: 91962961-91962961 on Build GRCh38
rs755333661 Chr.1: 91962839-91962839 on Build GRCh38
rs764562513 Chr.1: 91962943-91962943 on Build GRCh38
rs776641724 Chr.1: 91962967-91962970 on Build GRCh38
rs768707054 Chr.1: 91962893-91962893 on Build GRCh38
rs773548471 Chr.1: 91962988-91962990 on Build GRCh38

View Full Product Details