Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42374157-42374390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKK4
Entrez Gene ID
27121
Gene Name
dickkopf WNT signaling pathway inhibitor 4
Gene Aliases
DKK-4
UniGene
Hs.159311
Gene Chromosome Location
Chr.8: 42373194-42391322 on Build GRCh38
Transcripts
NM_014420.2 XM_011544488.2 XM_017013316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3900288 c.523G>C p.V175L Chr.8: 42374252-42374252 on Build GRCh38
3834747 c.578G>A p.R193H Chr.8: 42374197-42374197 on Build GRCh38
5962636 c.563C>T p.P188L Chr.8: 42374212-42374212 on Build GRCh38
3374932 c.416G>T p.G139V Chr.8: 42374359-42374359 on Build GRCh38
375129 c.468C>T p.C156C Chr.8: 42374307-42374307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899356263 Chr.8: 42374315-42374315 on Build GRCh38
rs982154368 Chr.8: 42374216-42374216 on Build GRCh38
rs199600734 Chr.8: 42374287-42374287 on Build GRCh38
rs752710793 Chr.8: 42374231-42374231 on Build GRCh38
rs777074997 Chr.8: 42374193-42374193 on Build GRCh38
rs779669823 Chr.8: 42374329-42374329 on Build GRCh38
rs541376818 Chr.8: 42374281-42374281 on Build GRCh38
rs761408532 Chr.8: 42374207-42374207 on Build GRCh38
rs765548766 Chr.8: 42374203-42374203 on Build GRCh38
rs764520274 Chr.8: 42374192-42374192 on Build GRCh38
rs750084403 Chr.8: 42374323-42374323 on Build GRCh38
rs969446163 Chr.8: 42374211-42374211 on Build GRCh38
rs771646549 Chr.8: 42374275-42374275 on Build GRCh38
rs35249735 Chr.8: 42374327-42374327 on Build GRCh38
rs747561014 Chr.8: 42374280-42374280 on Build GRCh38
rs529642073 Chr.8: 42374299-42374299 on Build GRCh38
rs755946609 Chr.8: 42374233-42374233 on Build GRCh38
rs758529240 Chr.8: 42374224-42374224 on Build GRCh38
rs753684391 Chr.8: 42374309-42374309 on Build GRCh38
rs905191716 Chr.8: 42374356-42374356 on Build GRCh38
rs2073664 Chr.8: 42374268-42374268 on Build GRCh38
rs750569149 Chr.8: 42374217-42374217 on Build GRCh38
rs774799806 Chr.8: 42374277-42374277 on Build GRCh38
rs372587994 Chr.8: 42374235-42374235 on Build GRCh38
rs1023926238 Chr.8: 42374341-42374341 on Build GRCh38
rs766319350 Chr.8: 42374229-42374229 on Build GRCh38
rs939765578 Chr.8: 42374290-42374290 on Build GRCh38
rs766494052 Chr.8: 42374300-42374300 on Build GRCh38
rs148982158 Chr.8: 42374238-42374238 on Build GRCh38
rs763989701 Chr.8: 42374306-42374306 on Build GRCh38
rs1036509058 Chr.8: 42374312-42374312 on Build GRCh38
rs908795687 Chr.8: 42374345-42374345 on Build GRCh38
rs765603868 Chr.8: 42374298-42374298 on Build GRCh38
rs375323773 Chr.8: 42374246-42374246 on Build GRCh38
rs756944574 Chr.8: 42374256-42374256 on Build GRCh38
rs757069150 Chr.8: 42374322-42374322 on Build GRCh38
rs1035109122 Chr.8: 42374225-42374225 on Build GRCh38
rs760734756 Chr.8: 42374305-42374305 on Build GRCh38
rs1024039764 Chr.8: 42374202-42374202 on Build GRCh38
rs115616321 Chr.8: 42374273-42374273 on Build GRCh38
rs114107982 Chr.8: 42374257-42374257 on Build GRCh38
rs777200031 Chr.8: 42374286-42374286 on Build GRCh38

View Full Product Details