Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64193926-64194187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf66
Entrez Gene ID
144577
Gene Name
chromosome 12 open reading frame 66
Gene Aliases
-
UniGene
Hs.444671
Gene Chromosome Location
Chr.12: 64186312-64222296 on Build GRCh38
Transcripts
NM_001300940.1 NM_001300941.1 NM_152440.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5734477 c.1231A>G p.K411E Chr.12: 64193949-64193949 on Build GRCh38
942467 c.1104G>A p.M368I Chr.12: 64194076-64194076 on Build GRCh38
3987148 c.1065C>T p.D355D Chr.12: 64194115-64194115 on Build GRCh38
4044140 c.1190G>A p.R397H Chr.12: 64193990-64193990 on Build GRCh38
6329745 c.1112G>T p.R371L Chr.12: 64194068-64194068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs180701944 Chr.12: 64194069-64194069 on Build GRCh38
rs746911314 Chr.12: 64194071-64194071 on Build GRCh38
rs571079002 Chr.12: 64194117-64194117 on Build GRCh38
rs779722376 Chr.12: 64194165-64194165 on Build GRCh38
rs755561216 Chr.12: 64193997-64193997 on Build GRCh38
rs776484826 Chr.12: 64194074-64194074 on Build GRCh38
rs777080708 Chr.12: 64194130-64194130 on Build GRCh38
rs753939407 Chr.12: 64194166-64194166 on Build GRCh38
rs766567978 Chr.12: 64193991-64193991 on Build GRCh38
rs761657897 Chr.12: 64194079-64194079 on Build GRCh38
rs758078738 Chr.12: 64194163-64194163 on Build GRCh38
rs771514284 Chr.12: 64194062-64194062 on Build GRCh38
rs752348517 Chr.12: 64194108-64194108 on Build GRCh38
rs750672936 Chr.12: 64193986-64193986 on Build GRCh38
rs769547546 Chr.12: 64194094-64194094 on Build GRCh38
rs758314142 Chr.12: 64194089-64194090 on Build GRCh38
rs1044598248 Chr.12: 64194158-64194158 on Build GRCh38
rs999001538 Chr.12: 64194037-64194037 on Build GRCh38
rs765477992 Chr.12: 64193972-64193972 on Build GRCh38
rs776442685 Chr.12: 64193957-64193957 on Build GRCh38
rs773421355 Chr.12: 64194104-64194104 on Build GRCh38
rs760365339 Chr.12: 64194004-64194004 on Build GRCh38
rs370394947 Chr.12: 64194118-64194118 on Build GRCh38
rs781653218 Chr.12: 64194001-64194001 on Build GRCh38
rs750044162 Chr.12: 64194148-64194148 on Build GRCh38
rs374731314 Chr.12: 64194110-64194110 on Build GRCh38
rs551606664 Chr.12: 64194132-64194132 on Build GRCh38
rs756496068 Chr.12: 64194137-64194137 on Build GRCh38
rs753072674 Chr.12: 64194017-64194017 on Build GRCh38
rs923638008 Chr.12: 64194007-64194007 on Build GRCh38
rs751718683 Chr.12: 64194113-64194113 on Build GRCh38
rs761556130 Chr.12: 64193970-64193970 on Build GRCh38
rs763086764 Chr.12: 64193987-64193987 on Build GRCh38
rs764369829 Chr.12: 64194139-64194139 on Build GRCh38
rs202171306 Chr.12: 64194048-64194048 on Build GRCh38
rs116379450 Chr.12: 64194115-64194115 on Build GRCh38
rs745573510 Chr.12: 64194052-64194052 on Build GRCh38
rs528981217 Chr.12: 64194022-64194022 on Build GRCh38
rs146776139 Chr.12: 64194073-64194073 on Build GRCh38

View Full Product Details