Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157311593-157311854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYFIP2
Entrez Gene ID
26999
Gene Name
cytoplasmic FMR1 interacting protein 2
Gene Aliases
PIR121
UniGene
Hs.519702
Gene Chromosome Location
Chr.5: 157266080-157395598 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3947187 c.44C>A p.P15H Chr.5: 157311693-157311693 on Build GRCh38
7363470 c.94C>T p.R32C Chr.5: 157311743-157311743 on Build GRCh38
5662555 c.130G>A p.E44K Chr.5: 157311779-157311779 on Build GRCh38
4521432 c.132G>C p.E44D Chr.5: 157311781-157311781 on Build GRCh38
6638858 c.61G>A p.E21K Chr.5: 157311710-157311710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376982797 Chr.5: 157311754-157311754 on Build GRCh38
rs760249068 Chr.5: 157311761-157311761 on Build GRCh38
rs547524850 Chr.5: 157311795-157311795 on Build GRCh38
rs370498979 Chr.5: 157311812-157311812 on Build GRCh38
rs745838142 Chr.5: 157311670-157311670 on Build GRCh38
rs926031238 Chr.5: 157311615-157311615 on Build GRCh38
rs548540530 Chr.5: 157311717-157311717 on Build GRCh38
rs776871043 Chr.5: 157311688-157311688 on Build GRCh38
rs376920120 Chr.5: 157311664-157311664 on Build GRCh38
rs148350606 Chr.5: 157311669-157311669 on Build GRCh38
rs1026289030 Chr.5: 157311626-157311626 on Build GRCh38
rs373676501 Chr.5: 157311712-157311712 on Build GRCh38
rs533425378 Chr.5: 157311620-157311620 on Build GRCh38
rs773461848 Chr.5: 157311619-157311619 on Build GRCh38
rs377619488 Chr.5: 157311641-157311641 on Build GRCh38
rs777561125 Chr.5: 157311646-157311646 on Build GRCh38
rs748234724 Chr.5: 157311823-157311823 on Build GRCh38
rs373707014 Chr.5: 157311649-157311649 on Build GRCh38
rs913123245 Chr.5: 157311788-157311788 on Build GRCh38
rs1051195028 Chr.5: 157311798-157311798 on Build GRCh38
rs780920901 Chr.5: 157311684-157311684 on Build GRCh38
rs10062193 Chr.5: 157311648-157311648 on Build GRCh38
rs370358417 Chr.5: 157311813-157311813 on Build GRCh38
rs752158612 Chr.5: 157311634-157311634 on Build GRCh38
rs373811779 Chr.5: 157311636-157311636 on Build GRCh38
rs577627376 Chr.5: 157311642-157311642 on Build GRCh38
rs758423007 Chr.5: 157311817-157311817 on Build GRCh38
rs763206176 Chr.5: 157311743-157311743 on Build GRCh38
rs771368440 Chr.5: 157311744-157311744 on Build GRCh38
rs752212345 Chr.5: 157311760-157311760 on Build GRCh38
rs1017783280 Chr.5: 157311709-157311709 on Build GRCh38
rs116166416 Chr.5: 157311789-157311789 on Build GRCh38
rs369031377 Chr.5: 157311628-157311628 on Build GRCh38
rs372848474 Chr.5: 157311724-157311724 on Build GRCh38
rs393178 Chr.5: 157311822-157311822 on Build GRCh38
rs568356129 Chr.5: 157311747-157311747 on Build GRCh38
rs760060931 Chr.5: 157311749-157311749 on Build GRCh38
rs748322670 Chr.5: 157311696-157311696 on Build GRCh38
rs1026182000 Chr.5: 157311769-157311769 on Build GRCh38
rs530666349 Chr.5: 157311640-157311640 on Build GRCh38
rs766837461 Chr.5: 157311808-157311814 on Build GRCh38
rs765647472 Chr.5: 157311618-157311618 on Build GRCh38
rs990110659 Chr.5: 157311729-157311729 on Build GRCh38

View Full Product Details