Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49704979-49705220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPT1C
Entrez Gene ID
126129
Gene Name
carnitine palmitoyltransferase 1C
Gene Aliases
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
UniGene
Hs.112195
Gene Chromosome Location
Chr.19: 49690672-49713731 on Build GRCh38
Transcripts
NM_001136052.2 NM_001199752.2 NM_001199753.1 NM_152359.2 NR_108072.1 XM_005258505.2 XM_005258506.4 XM_006723009.3 XM_011526438.2 XM_011526439.2 XM_011526440.2 XM_017026265.1 XM_017026266.1 XM_017026267.1 XM_017026268.1 XM_017026269.1 XM_017026270.1 XM_017026271.1 XM_017026272.1 XM_017026273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1743087 c.886C>A p.L296M Chr.19: 49705220-49705220 on Build GRCh38
3893066 c.825C>T p.A275A Chr.19: 49705060-49705060 on Build GRCh38
3893067 c.841C>T p.L281L Chr.19: 49705076-49705076 on Build GRCh38
4674499 c.815C>A p.A272D Chr.19: 49705050-49705050 on Build GRCh38
4674498 c.812G>A p.R271H Chr.19: 49705047-49705047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024589941 Chr.19: 49705102-49705102 on Build GRCh38
rs772025553 Chr.19: 49705083-49705083 on Build GRCh38
rs201458521 Chr.19: 49705046-49705046 on Build GRCh38
rs1049933458 Chr.19: 49705114-49705114 on Build GRCh38
rs770092727 Chr.19: 49705015-49705015 on Build GRCh38
rs761167173 Chr.19: 49705100-49705100 on Build GRCh38
rs754172418 Chr.19: 49705116-49705116 on Build GRCh38
rs755823050 Chr.19: 49705103-49705104 on Build GRCh38
rs756866946 Chr.19: 49705075-49705075 on Build GRCh38
rs745328410 Chr.19: 49705082-49705082 on Build GRCh38
rs747927748 Chr.19: 49705171-49705171 on Build GRCh38
rs903998196 Chr.19: 49705025-49705025 on Build GRCh38
rs764369739 Chr.19: 49705113-49705113 on Build GRCh38
rs370379621 Chr.19: 49705165-49705165 on Build GRCh38
rs774595895 Chr.19: 49705097-49705097 on Build GRCh38
rs771935586 Chr.19: 49705005-49705005 on Build GRCh38
rs1055311456 Chr.19: 49705088-49705088 on Build GRCh38
rs1042583182 Chr.19: 49705014-49705014 on Build GRCh38
rs368063438 Chr.19: 49705146-49705146 on Build GRCh38
rs753427726 Chr.19: 49705063-49705063 on Build GRCh38
rs772272448 Chr.19: 49705196-49705196 on Build GRCh38
rs763153595 Chr.19: 49705029-49705029 on Build GRCh38
rs764826051 Chr.19: 49705128-49705128 on Build GRCh38
rs746873874 Chr.19: 49705085-49705085 on Build GRCh38
rs749973168 Chr.19: 49705130-49705130 on Build GRCh38
rs373790571 Chr.19: 49705172-49705172 on Build GRCh38
rs779932649 Chr.19: 49705084-49705084 on Build GRCh38
rs768088467 Chr.19: 49705047-49705047 on Build GRCh38
rs745809920 Chr.19: 49705004-49705004 on Build GRCh38
rs199544289 Chr.19: 49705030-49705030 on Build GRCh38
rs200717435 Chr.19: 49705048-49705048 on Build GRCh38
rs375077196 Chr.19: 49705143-49705143 on Build GRCh38
rs745995686 Chr.19: 49705194-49705194 on Build GRCh38
rs763094054 Chr.19: 49705089-49705089 on Build GRCh38
rs143361095 Chr.19: 49705098-49705098 on Build GRCh38
rs367605898 Chr.19: 49705034-49705034 on Build GRCh38
rs777538460 Chr.19: 49705060-49705060 on Build GRCh38
rs756921758 Chr.19: 49705118-49705118 on Build GRCh38
rs140099471 Chr.19: 49705090-49705090 on Build GRCh38
rs773288049 Chr.19: 49705021-49705021 on Build GRCh38
rs768326297 Chr.19: 49705087-49705087 on Build GRCh38
rs756410814 Chr.19: 49705049-49705049 on Build GRCh38
rs747560763 Chr.19: 49705126-49705126 on Build GRCh38
rs947674908 Chr.19: 49705110-49705110 on Build GRCh38
rs368773100 Chr.19: 49705182-49705182 on Build GRCh38
rs200395771 Chr.19: 49705009-49705009 on Build GRCh38
rs889954627 Chr.19: 49705127-49705127 on Build GRCh38
rs760344623 Chr.19: 49705010-49705010 on Build GRCh38
rs780442933 Chr.19: 49705108-49705109 on Build GRCh38

View Full Product Details