Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68422334-68422597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNA2
Entrez Gene ID
1763
Gene Name
DNA replication helicase/nuclease 2
Gene Aliases
DNA2L, hDNA2
UniGene
Hs.532446
Gene Chromosome Location
Chr.10: 68414064-68472521 on Build GRCh38
Transcripts
NM_001080449.2 NR_102264.1 XM_006717680.3 XM_011539417.1 XM_017015799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6895053 c.2487G>A p.M829I Chr.10: 68422520-68422520 on Build GRCh38
1474715 c.2414T>G p.M805R Chr.10: 68422593-68422593 on Build GRCh38
3439862 c.2794C>A p.L932M Chr.10: 68422386-68422386 on Build GRCh38
1474714 c.2672T>G p.M891R Chr.10: 68422593-68422593 on Build GRCh38
6845508 c.2770A>T p.N924Y Chr.10: 68422410-68422410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777394634 Chr.10: 68422560-68422560 on Build GRCh38
rs983210057 Chr.10: 68422508-68422508 on Build GRCh38
rs753500591 Chr.10: 68422506-68422507 on Build GRCh38
rs574169301 Chr.10: 68422400-68422400 on Build GRCh38
rs761979661 Chr.10: 68422477-68422477 on Build GRCh38
rs1027322616 Chr.10: 68422472-68422472 on Build GRCh38
rs751612648 Chr.10: 68422392-68422392 on Build GRCh38
rs193299918 Chr.10: 68422433-68422433 on Build GRCh38
rs761138552 Chr.10: 68422455-68422455 on Build GRCh38
rs755953324 Chr.10: 68422544-68422544 on Build GRCh38
rs772653603 Chr.10: 68422448-68422448 on Build GRCh38
rs767523892 Chr.10: 68422535-68422535 on Build GRCh38
rs543180602 Chr.10: 68422420-68422420 on Build GRCh38
rs370020989 Chr.10: 68422487-68422487 on Build GRCh38
rs752403785 Chr.10: 68422539-68422539 on Build GRCh38
rs746296708 Chr.10: 68422444-68422444 on Build GRCh38
rs764385094 Chr.10: 68422464-68422464 on Build GRCh38
rs754945524 Chr.10: 68422395-68422395 on Build GRCh38
rs959351316 Chr.10: 68422364-68422364 on Build GRCh38
rs2273904 Chr.10: 68422511-68422511 on Build GRCh38
rs372971290 Chr.10: 68422418-68422418 on Build GRCh38
rs563046954 Chr.10: 68422428-68422428 on Build GRCh38
rs777050835 Chr.10: 68422470-68422470 on Build GRCh38
rs201513130 Chr.10: 68422534-68422534 on Build GRCh38
rs775674923 Chr.10: 68422451-68422451 on Build GRCh38
rs994876466 Chr.10: 68422449-68422449 on Build GRCh38
rs375912198 Chr.10: 68422429-68422429 on Build GRCh38

View Full Product Details