Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 54962854-54963091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX11
Entrez Gene ID
1353
Gene Name
COX11, cytochrome c oxidase copper chaperone
Gene Aliases
COX11P
UniGene
Hs.591171
Gene Chromosome Location
Chr.17: 54951898-54971024 on Build GRCh38
Transcripts
NM_001162861.2 NM_001162862.2 NM_001321518.1 NM_004375.4 NR_027941.2 NR_027942.2 NR_135677.1 XM_011524342.2 XM_017024192.1 XM_017024193.1 XM_017024194.1 XM_017024195.1 XM_017024196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5553056 c.661G>A p.E221K Chr.17: 54962903-54962903 on Build GRCh38
7276041 c.688G>A p.E230K Chr.17: 54962876-54962876 on Build GRCh38
5563444 c.682C>A p.Q228K Chr.17: 54962882-54962882 on Build GRCh38
389212 c.654C>T p.F218F Chr.17: 54962910-54962910 on Build GRCh38
4416836 c.698T>C p.M233T Chr.17: 54962866-54962866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756362909 Chr.17: 54962881-54962881 on Build GRCh38
rs557091152 Chr.17: 54963041-54963041 on Build GRCh38
rs758304825 Chr.17: 54963028-54963028 on Build GRCh38
rs779471204 Chr.17: 54962926-54962926 on Build GRCh38
rs767566840 Chr.17: 54963038-54963039 on Build GRCh38
rs774294644 Chr.17: 54962986-54962990 on Build GRCh38
rs764408289 Chr.17: 54962887-54962887 on Build GRCh38
rs140907287 Chr.17: 54962956-54962956 on Build GRCh38
rs376571839 Chr.17: 54962908-54962908 on Build GRCh38
rs759476892 Chr.17: 54962968-54962969 on Build GRCh38
rs780636122 Chr.17: 54962948-54962948 on Build GRCh38
rs369696146 Chr.17: 54962977-54962977 on Build GRCh38
rs917042839 Chr.17: 54962986-54962986 on Build GRCh38
rs754500625 Chr.17: 54962939-54962939 on Build GRCh38
rs746312332 Chr.17: 54962938-54962938 on Build GRCh38
rs769412588 Chr.17: 54962963-54962963 on Build GRCh38
rs757762886 Chr.17: 54962919-54962919 on Build GRCh38
rs201010092 Chr.17: 54962892-54962892 on Build GRCh38
rs949830540 Chr.17: 54963010-54963010 on Build GRCh38
rs560397238 Chr.17: 54963039-54963039 on Build GRCh38

View Full Product Details