Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 79680135-79680382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDP1
Entrez Gene ID
9150
Gene Name
CTD phosphatase subunit 1
Gene Aliases
CCFDN, FCP1
UniGene
Hs.465490
Gene Chromosome Location
Chr.18: 79679801-79754510 on Build GRCh38
Transcripts
NM_001318511.1 NM_004715.4 NM_048368.3 XM_011526261.1 XM_011526290.2 XM_017026112.1

Gene Symbol
LOC105376875
Entrez Gene ID
105376875
Gene Name
uncharacterized LOC105376875
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.18: 79680284-79688319 on Build GRCh38
Transcripts
NM_001318511.1 NM_004715.4 NM_048368.3 XM_011526261.1 XM_011526290.2 XM_017026112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3766260 c.213G>C p.G71G Chr.18: 79680160-79680160 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1028182398 Chr.18: 79680163-79680163 on Build GRCh38
rs898359609 Chr.18: 79680277-79680277 on Build GRCh38
rs1021090497 Chr.18: 79680214-79680214 on Build GRCh38
rs553405714 Chr.18: 79680285-79680285 on Build GRCh38
rs940048700 Chr.18: 79680327-79680327 on Build GRCh38
rs984222178 Chr.18: 79680305-79680305 on Build GRCh38
rs201234471 Chr.18: 79680160-79680160 on Build GRCh38
rs1035960838 Chr.18: 79680242-79680242 on Build GRCh38
rs893643197 Chr.18: 79680178-79680178 on Build GRCh38
rs770292192 Chr.18: 79680169-79680169 on Build GRCh38
rs551929556 Chr.18: 79680337-79680337 on Build GRCh38
rs73972710 Chr.18: 79680324-79680324 on Build GRCh38
rs187756055 Chr.18: 79680241-79680241 on Build GRCh38
rs967660848 Chr.18: 79680351-79680351 on Build GRCh38
rs960310495 Chr.18: 79680264-79680264 on Build GRCh38
rs949746367 Chr.18: 79680212-79680212 on Build GRCh38
rs936183704 Chr.18: 79680167-79680167 on Build GRCh38
rs768583929 Chr.18: 79680207-79680207 on Build GRCh38
rs915649307 Chr.18: 79680201-79680201 on Build GRCh38
rs113762153 Chr.18: 79680361-79680361 on Build GRCh38
rs533419347 Chr.18: 79680326-79680326 on Build GRCh38
rs774697090 Chr.18: 79680227-79680227 on Build GRCh38
rs769337405 Chr.18: 79680314-79680314 on Build GRCh38
rs932595680 Chr.18: 79680342-79680342 on Build GRCh38
rs1049665889 Chr.18: 79680347-79680347 on Build GRCh38
rs966405383 Chr.18: 79680222-79680222 on Build GRCh38
rs763118972 Chr.18: 79680188-79680188 on Build GRCh38
rs1038208260 Chr.18: 79680246-79680246 on Build GRCh38
rs771395058 Chr.18: 79680153-79680153 on Build GRCh38
rs931445557 Chr.18: 79680299-79680299 on Build GRCh38
rs111853095 Chr.18: 79680164-79680164 on Build GRCh38
rs534987615 Chr.18: 79680256-79680256 on Build GRCh38
rs996752979 Chr.18: 79680162-79680162 on Build GRCh38
rs991697139 Chr.18: 79680282-79680282 on Build GRCh38
rs1040984549 Chr.18: 79680156-79680156 on Build GRCh38
rs887043576 Chr.18: 79680232-79680232 on Build GRCh38
rs996672871 Chr.18: 79680281-79680281 on Build GRCh38
rs901153928 Chr.18: 79680341-79680341 on Build GRCh38
rs775939813 Chr.18: 79680172-79680172 on Build GRCh38
rs112059691 Chr.18: 79680206-79680206 on Build GRCh38

View Full Product Details