Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40021442-40021711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83F
Entrez Gene ID
113828
Gene Name
family with sequence similarity 83 member F
Gene Aliases
-
UniGene
Hs.197680
Gene Chromosome Location
Chr.22: 39994949-40030039 on Build GRCh38
Transcripts
NM_138435.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6660235 c.974C>A p.P325H Chr.22: 40021484-40021484 on Build GRCh38
4683255 c.1156delC p.V388fs*9 Chr.22: 40021666-40021666 on Build GRCh38
231450 c.1051G>A p.A351T Chr.22: 40021561-40021561 on Build GRCh38
3694067 c.1000C>T p.R334C Chr.22: 40021510-40021510 on Build GRCh38
6660237 c.1173delC p.L393fs*4 Chr.22: 40021683-40021683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761699833 Chr.22: 40021628-40021628 on Build GRCh38
rs34509121 Chr.22: 40021555-40021556 on Build GRCh38
rs902856112 Chr.22: 40021677-40021677 on Build GRCh38
rs746472470 Chr.22: 40021686-40021686 on Build GRCh38
rs761893807 Chr.22: 40021574-40021574 on Build GRCh38
rs762463384 Chr.22: 40021473-40021473 on Build GRCh38
rs145137330 Chr.22: 40021599-40021599 on Build GRCh38
rs11914082 Chr.22: 40021491-40021491 on Build GRCh38
rs759793024 Chr.22: 40021659-40021659 on Build GRCh38
rs373307669 Chr.22: 40021611-40021611 on Build GRCh38
rs1049561597 Chr.22: 40021656-40021656 on Build GRCh38
rs747373194 Chr.22: 40021464-40021464 on Build GRCh38
rs753758914 Chr.22: 40021517-40021517 on Build GRCh38
rs201378321 Chr.22: 40021547-40021547 on Build GRCh38
rs145428025 Chr.22: 40021546-40021546 on Build GRCh38
rs746620839 Chr.22: 40021612-40021612 on Build GRCh38
rs17850725 Chr.22: 40021555-40021555 on Build GRCh38
rs753117885 Chr.22: 40021669-40021669 on Build GRCh38
rs760501103 Chr.22: 40021511-40021511 on Build GRCh38
rs1020362697 Chr.22: 40021666-40021666 on Build GRCh38
rs140734376 Chr.22: 40021492-40021492 on Build GRCh38
rs758495757 Chr.22: 40021594-40021594 on Build GRCh38
rs932840488 Chr.22: 40021617-40021617 on Build GRCh38
rs1021836776 Chr.22: 40021553-40021553 on Build GRCh38
rs750538925 Chr.22: 40021587-40021587 on Build GRCh38
rs1009859953 Chr.22: 40021635-40021635 on Build GRCh38
rs369369585 Chr.22: 40021610-40021610 on Build GRCh38
rs781668146 Chr.22: 40021551-40021551 on Build GRCh38
rs763939183 Chr.22: 40021566-40021566 on Build GRCh38
rs769113395 Chr.22: 40021466-40021466 on Build GRCh38
rs776961253 Chr.22: 40021671-40021671 on Build GRCh38
rs551543842 Chr.22: 40021650-40021650 on Build GRCh38
rs768403243 Chr.22: 40021618-40021618 on Build GRCh38
rs573958791 Chr.22: 40021575-40021575 on Build GRCh38
rs765362025 Chr.22: 40021535-40021535 on Build GRCh38
rs759158719 Chr.22: 40021498-40021498 on Build GRCh38
rs557259135 Chr.22: 40021543-40021543 on Build GRCh38
rs375841212 Chr.22: 40021556-40021556 on Build GRCh38
rs1047489111 Chr.22: 40021593-40021593 on Build GRCh38
rs763947933 Chr.22: 40021515-40021515 on Build GRCh38
rs114194620 Chr.22: 40021604-40021604 on Build GRCh38
rs769914565 Chr.22: 40021639-40021639 on Build GRCh38
rs767721247 Chr.22: 40021660-40021660 on Build GRCh38
rs375725780 Chr.22: 40021649-40021649 on Build GRCh38
rs778239530 Chr.22: 40021609-40021609 on Build GRCh38
rs199712785 Chr.22: 40021606-40021606 on Build GRCh38
rs768492903 Chr.22: 40021691-40021691 on Build GRCh38
rs931272926 Chr.22: 40021585-40021585 on Build GRCh38
rs745378535 Chr.22: 40021558-40021558 on Build GRCh38
rs533312678 Chr.22: 40021619-40021619 on Build GRCh38
rs149665361 Chr.22: 40021510-40021510 on Build GRCh38
rs770937456 Chr.22: 40021534-40021534 on Build GRCh38
rs755313030 Chr.22: 40021549-40021549 on Build GRCh38
rs748615953 Chr.22: 40021552-40021552 on Build GRCh38
rs777110834 Chr.22: 40021469-40021469 on Build GRCh38
rs760387598 Chr.22: 40021563-40021563 on Build GRCh38
rs981221915 Chr.22: 40021520-40021520 on Build GRCh38
rs757440995 Chr.22: 40021518-40021518 on Build GRCh38
rs202183455 Chr.22: 40021580-40021581 on Build GRCh38
rs35823589 Chr.22: 40021567-40021567 on Build GRCh38
rs1008750037 Chr.22: 40021502-40021502 on Build GRCh38
rs775146771 Chr.22: 40021562-40021562 on Build GRCh38
rs773144944 Chr.22: 40021642-40021642 on Build GRCh38
rs754327344 Chr.22: 40021672-40021672 on Build GRCh38
rs756679205 Chr.22: 40021608-40021608 on Build GRCh38
rs747392064 Chr.22: 40021548-40021548 on Build GRCh38
rs755330657 Chr.22: 40021602-40021602 on Build GRCh38
rs138973979 Chr.22: 40021600-40021600 on Build GRCh38
rs140645542 Chr.22: 40021679-40021679 on Build GRCh38
rs961050448 Chr.22: 40021605-40021605 on Build GRCh38

View Full Product Details