Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27726008-27726237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM76A
Entrez Gene ID
199870
Gene Name
family with sequence similarity 76 member A
Gene Aliases
-
UniGene
Hs.469359
Gene Chromosome Location
Chr.1: 27725979-27762915 on Build GRCh38
Transcripts
NM_001143912.1 NM_001143913.1 NM_001143914.1 NM_001143915.1 NM_152660.2 XM_017000543.1

SNP Detail

SNP ID Chromosome Location
rs866249242 Chr.1: 27726153-27726153 on Build GRCh38
rs867203074 Chr.1: 27726144-27726144 on Build GRCh38
rs534833333 Chr.1: 27726186-27726186 on Build GRCh38
rs555276308 Chr.1: 27726165-27726165 on Build GRCh38
rs769392735 Chr.1: 27726145-27726145 on Build GRCh38
rs753895701 Chr.1: 27726209-27726209 on Build GRCh38
rs1035939334 Chr.1: 27726052-27726052 on Build GRCh38
rs774802281 Chr.1: 27726171-27726171 on Build GRCh38
rs866711858 Chr.1: 27726157-27726157 on Build GRCh38
rs867590598 Chr.1: 27726152-27726152 on Build GRCh38
rs764185395 Chr.1: 27726208-27726208 on Build GRCh38
rs867411986 Chr.1: 27726141-27726141 on Build GRCh38
rs1049049196 Chr.1: 27726036-27726036 on Build GRCh38
rs756126896 Chr.1: 27726191-27726191 on Build GRCh38
rs567370122 Chr.1: 27726077-27726077 on Build GRCh38
rs1034773600 Chr.1: 27726215-27726215 on Build GRCh38
rs535352798 Chr.1: 27726146-27726146 on Build GRCh38
rs565510183 Chr.1: 27726180-27726180 on Build GRCh38
rs1001904398 Chr.1: 27726184-27726184 on Build GRCh38
rs753005255 Chr.1: 27726187-27726187 on Build GRCh38
rs887329420 Chr.1: 27726043-27726043 on Build GRCh38
rs747578182 Chr.1: 27726119-27726119 on Build GRCh38
rs866228830 Chr.1: 27726142-27726142 on Build GRCh38
rs776409525 Chr.1: 27726070-27726070 on Build GRCh38
rs1004420325 Chr.1: 27726044-27726044 on Build GRCh38
rs768283800 Chr.1: 27726056-27726056 on Build GRCh38
rs969557419 Chr.1: 27726192-27726192 on Build GRCh38
rs995657904 Chr.1: 27726035-27726035 on Build GRCh38

View Full Product Details