Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128838384-128838633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNC
Entrez Gene ID
2318
Gene Name
filamin C
Gene Aliases
ABP-280, ABP280A, ABPA, ABPL, FLN2, MFM5, MPD4
UniGene
Hs.58414
Gene Chromosome Location
Chr.7: 128830429-128859274 on Build GRCh38
Transcripts
NM_001127487.1 NM_001458.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6299213 c.1211-5G>A p.? Chr.7: 128838598-128838598 on Build GRCh38
4684989 c.1225G>A p.D409N Chr.7: 128838617-128838617 on Build GRCh38
2769308 c.1224C>T p.G408G Chr.7: 128838616-128838616 on Build GRCh38
6176228 c.1235T>C p.V412A Chr.7: 128838627-128838627 on Build GRCh38
6108682 c.1217G>A p.G406D Chr.7: 128838609-128838609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750881912 Chr.7: 128838403-128838403 on Build GRCh38
rs766697953 Chr.7: 128838553-128838553 on Build GRCh38
rs756772848 Chr.7: 128838578-128838578 on Build GRCh38
rs778565971 Chr.7: 128838419-128838419 on Build GRCh38
rs925579720 Chr.7: 128838484-128838484 on Build GRCh38
rs751006412 Chr.7: 128838577-128838577 on Build GRCh38
rs776554557 Chr.7: 128838436-128838436 on Build GRCh38
rs771914549 Chr.7: 128838492-128838492 on Build GRCh38
rs762402939 Chr.7: 128838475-128838475 on Build GRCh38
rs761064939 Chr.7: 128838552-128838552 on Build GRCh38
rs770923888 Chr.7: 128838434-128838434 on Build GRCh38
rs768065942 Chr.7: 128838477-128838477 on Build GRCh38
rs750976968 Chr.7: 128838479-128838479 on Build GRCh38
rs774628429 Chr.7: 128838465-128838465 on Build GRCh38
rs777199447 Chr.7: 128838427-128838427 on Build GRCh38
rs374757755 Chr.7: 128838424-128838424 on Build GRCh38
rs757898362 Chr.7: 128838425-128838425 on Build GRCh38
rs967225350 Chr.7: 128838568-128838568 on Build GRCh38
rs756546984 Chr.7: 128838416-128838416 on Build GRCh38
rs749510534 Chr.7: 128838607-128838607 on Build GRCh38
rs906565119 Chr.7: 128838493-128838493 on Build GRCh38
rs562941732 Chr.7: 128838439-128838439 on Build GRCh38
rs368892134 Chr.7: 128838437-128838437 on Build GRCh38
rs575934150 Chr.7: 128838486-128838486 on Build GRCh38
rs769599454 Chr.7: 128838598-128838598 on Build GRCh38
rs894370482 Chr.7: 128838535-128838535 on Build GRCh38
rs547183303 Chr.7: 128838555-128838555 on Build GRCh38
rs779760488 Chr.7: 128838599-128838599 on Build GRCh38
rs113882013 Chr.7: 128838506-128838506 on Build GRCh38
rs1002168964 Chr.7: 128838500-128838500 on Build GRCh38
rs746938160 Chr.7: 128838428-128838428 on Build GRCh38
rs745834917 Chr.7: 128838597-128838597 on Build GRCh38
rs768757318 Chr.7: 128838460-128838460 on Build GRCh38
rs763122529 Chr.7: 128838444-128838444 on Build GRCh38
rs113902193 Chr.7: 128838551-128838551 on Build GRCh38
rs1050963705 Chr.7: 128838539-128838539 on Build GRCh38
rs35456196 Chr.7: 128838539-128838539 on Build GRCh38
rs775741829 Chr.7: 128838442-128838442 on Build GRCh38
rs933479131 Chr.7: 128838527-128838527 on Build GRCh38
rs777333499 Chr.7: 128838571-128838571 on Build GRCh38
rs368219607 Chr.7: 128838558-128838558 on Build GRCh38
rs546166999 Chr.7: 128838494-128838494 on Build GRCh38
rs781059433 Chr.7: 128838585-128838585 on Build GRCh38
rs1016025623 Chr.7: 128838495-128838495 on Build GRCh38
rs368590071 Chr.7: 128838525-128838525 on Build GRCh38
rs978392380 Chr.7: 128838438-128838438 on Build GRCh38

View Full Product Details